Skip to Content
Merck
All Photos(2)

Key Documents

91822

Supelco

5-Pentadecylresorcinol

analytical standard

Synonym(s):

1,3-Dihydroxy-5-pentadecylbenzene, 5-Pentadecyl-1,3-benzenediol

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$882.00

$882.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$882.00

About This Item

Empirical Formula (Hill Notation):
C21H36O2
CAS Number:
Molecular Weight:
320.51
Beilstein:
1982742
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

$882.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCCCCc1cc(O)cc(O)c1

InChI

1S/C21H36O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-19-16-20(22)18-21(23)17-19/h16-18,22-23H,2-15H2,1H3

InChI key

KVVSCMOUFCNCGX-UHFFFAOYSA-N

General description

5-Pentadecylresorcinol is a resorcinol derivative[1], that can be found in the wood of Grevillea.banksii.[2]
Occurs in peel and flesh of unripe mangos, Mangifera indica (Anacardiaceae).

Application

5-Pentadecylresorcinol has been used as an analytical reference standard for the quantification of the analyte in a mixture of alkylresorcinols using liquid chromatography coupled to mass spectrometry (LC-MS). It may also be used as an analytical reference standard for the determination of the analyte in:
  • Goji berries using ultra-high-pressure liquid chromatography coupled with quadrupole-time-of-flight mass spectrometry (UHPLC-ESI-QTOF-MS).[3]
  • Mango (Mangifera indica L.) peels using high-performance liquid chromatography/atmospheric pressure chemical ionization mass spectrometry (HPLC/APCI-MS).[4]

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Deszcz et al.
Biochimica et biophysica acta, 1483(2), 241-250 (2000-01-15)
The occurrence of alkylresorcinols, polyketide compounds that in the same homologous series as cardol isolated from Anacardium occidentale (cashew) or bilobol from Ginkgo biloba which are derivatives of 1,3-dihydroxy-5-alk(en)ylbenzene, have been demonstrated in developing rye (Secale cereale L.) kernels. The
Ta-Hsien Chuang et al.
Journal of natural products, 70(2), 319-323 (2007-01-25)
Bioassay-guided fractionation of the MeOH extract of the leaves of Grevillea robusta led to the isolation of six new 5-alkylresorcinols, gravicycle (1), dehydrogravicycle (2), bisgravillol (3), dehydrobisgravillol (4), dehydrograviphane (5), and methyldehydrograviphane (6), as well as eight known compounds. The
J G Marks et al.
Journal of the American Academy of Dermatology, 10(4), 627-631 (1984-04-01)
Between April 4 and May 10, 1982, fifty-four individuals developed a poison ivy-like dermatitis 1 to 8 days after eating imported cashew nuts. The patients had a very pruritic, erythematous, maculopapular eruption that was accentuated in the flexural areas of
Spoerke and Smolinske.CS et al.
Toxicity of Houseplants (1990)
N Tsuge et al.
The Journal of antibiotics, 45(6), 886-891 (1992-06-01)
As a result of screening for inhibitors of glycerol-3-phosphate dehydrogenase, which may be effective to prevent corpulence, we isolated two inhibitors named adipostatin A and adipostatin B from the culture broth of Streptomyces cyaneus 2299-SV1. Their structures have been established

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service