Skip to Content
Merck
All Photos(1)

Key Documents

SMB00025

Sigma-Aldrich

Aspterric acid

≥95% (LC/MS-ELSD)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$938.00

$938.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
1 MG
$938.00

About This Item

Empirical Formula (Hill Notation):
C15H22O4
CAS Number:
Molecular Weight:
266.33
UNSPSC Code:
12352205
NACRES:
NA.25

$938.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 2 weeks.

Request a Bulk Order

Assay

≥95% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O1[C@H]2[C@@H](CC[C@@H]3C(C1)(C2)CCC3=C(C)C)C(=O)OO

InChI

1S/C15H22O4/c1-9(2)10-5-6-15-7-13(18-8-15)11(14(16)19-17)3-4-12(10)15/h11-13,17H,3-8H2,1-2H3/t11-,12+,13-,15?/m1/s1

InChI key

AMVWWAIZPPESMM-WORDMNGFSA-N

General description

Natural product derived from fungal source.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Atsumi Shimada et al.
Zeitschrift fur Naturforschung. C, Journal of biosciences, 63(7-8), 554-556 (2008-09-25)
Application of 100 microM aspterric acid (AA), a pollen growth inhibitor, with different concentrations of indole-3-acetic acid (IAA) results in the recovery of normal pollen development of Arabidopsis thaliana. Treatment with 100 microM AA plus 5 mM IAA significantly induced
Atsumi Shimada et al.
Zeitschrift fur Naturforschung. C, Journal of biosciences, 57(5-6), 459-464 (2002-07-23)
Aspterric Acid, 6-Hydroxymellein, Arabidopsis thaliana, Aspergillus terreus Aspterric acid (1) and 6-hydroxymellein (2), inhibitors of pollen development in Arabidopsis thaliana, have been isolated from the fungus Aspergillus terreus. 1 and 2 inhibited the pollen development at concentrations of 38 and
Atsumi Shimada et al.
Zeitschrift fur Naturforschung. C, Journal of biosciences, 60(7-8), 572-576 (2005-09-17)
Application of indole-3-acetic acid (IAA) with a pollen growth inhibitor, aspterric acid (AA), results in the recovery of normal pollen development. In contrast, application of gibberellin (GA3) with AA do not induce normal pollen growth. In addition, application of different

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service