Skip to Content
Merck
All Photos(1)

Documents

LM6005

Avanti

Cer/Sph Mixture II

Avanti Research - A Croda Brand

Synonym(s):

Cer/Sph Mixture II, Ceramide/Sphingoid Internal Standard Mixture II

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.25

form

ethanol solution

packaging

pkg of 1 × 1 mL (LM6005-1EA)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

~10 μg/mL (Refer to C of A for lot specific concentration. )

application(s)

lipidomics
metabolomics

shipped in

dry ice

storage temp.

−20°C

Application

Cer/Sph Mixture II has been used: as an internal standard mixture for sphingolipids quantification by liquid chromatography electrospray ionization-tandem mass spectrometry (LC ESI-MS/MS), in liquid chromatography-mass spectrometry (LC-MS) analysis for quality control check on the instrumental stability, internal standards mix for lipid extraction from serum and tissue samples by methyl-tert-butyl ether (MTBE)/methanol extraction method.

Packaging

2 mL Amber Glass Sealed Ampule (LM6005-1EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F

Flash Point(C)

13 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diego Arango et al.
The American journal of pathology, 180(4), 1509-1521 (2012-02-22)
Colorectal cancers (CRCs) are classified as having microsatellite instability (MSI) or chromosomal instability (CIN); herein termed microsatellite stable (MSS). MSI colon cancers frequently display a poorly differentiated histology for which the molecular basis is not well understood. Gene expression and
Merja Joensuu et al.
Progress in neuro-psychopharmacology & biological psychiatry, 84(Pt B), 362-381 (2017-09-06)
Dendritic spines are small actin-rich protrusions from neuronal dendrites that form the postsynaptic part of most excitatory synapses. Changes in the shape and size of dendritic spines correlate with the functional changes in excitatory synapses and are heavily dependent on
Jeffrey Molendijk et al.
Biomolecules, 10(5) (2020-05-21)
Esophageal adenocarcinoma (EAC) incidence has been rapidly increasing, potentially associated with the prevalence of the risk factors gastroesophageal reflux disease (GERD), obesity, high-fat diet (HFD), and the precursor condition Barrett's esophagus (BE). EAC development occurs over several years, with stepwise
Seema Khurana et al.
FEBS letters, 582(14), 2128-2139 (2008-03-01)
Villin is a tissue-specific actin modifying protein that is associated with actin filaments in the microvilli and terminal web of epithelial cells. It belongs to a large family of actin-binding proteins which includes actin-capping, -nucleating and/or -severing proteins such as
Nitai C Hait et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 34(3), 4293-4310 (2020-02-06)
Sphingosine kinase 2 (SphK2) is known to phosphorylate the nuclear sphingolipid metabolite to generate sphingosine-1-phosphate (S1P). Nuclear S1P is involved in epigenetic regulation of gene expression; however, the underlying mechanisms are not well understood. In this work, we have identified

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service