Key Documents
SLMTBP5-EP
Eppendorf® Safe-Lock Microcentrifuge Tubes, Biopur®
volume 0.5 mL, colorless, pkg of 50 ea (individually wrapped)
About This Item
UNSPSC Code:
41121703
Pricing and availability is not currently available.
sterility
sterile
packaging
pkg of 50 ea (individually wrapped)
manufacturer/tradename
Eppendorf® 022600001 (North America)
volume
0.5 mL
color
colorless
Related Categories
General description
  • Patented Safe-Lock hinged lid that eliminates any danger from leaks or from opening accidentally
  • Can be autoclaved when open (121°C, 20 minutes)
  • Has volume graduations and etched marking surfaces
  • Individually sealed Biopur microcentrifuge tube is sterile and free of pyrogens, RNase, DNA and ATP
Legal Information
Biopur is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)