Skip to Content
Merck
All Photos(1)

Key Documents

SCP0117

Sigma-Aldrich

Cionin Peptide

≥95% (HPLC), lyophilized powder

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
$279.00
1 MG
$434.00

$279.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 2 weeks.


Select a Size

Change View
500 μG
$279.00
1 MG
$434.00

About This Item

Empirical Formula (Hill Notation):
C53H63N11O19S3
Molecular Weight:
1254.32
UNSPSC Code:
12352202
NACRES:
NA.32

$279.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 2 weeks.

Product Name

Cionin,

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥85%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Asn-Tyr(SO3H)-Tyr(SO3H)-Gly-Trp-Met-Asp-Phe-NH2

Application

Protochordean octapeptide cionin is a structural hybrid of mammalian cholecystokinin (CCK) and gastrin with functions most similar to those of CCK. Cionin is used as a potent and efficient stimulator of gastric histamine release and acid secretion via a CCK-B receptor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stimulation by the ancestral member of the CCK/gastrin family, cionin, of trout gallbladder contraction.
A H Johnsen et al.
Annals of the New York Academy of Sciences, 713, 404-406 (1994-03-23)
Tsuyoshi Kawada et al.
Zoological science, 27(2), 134-153 (2010-02-10)
The critical phylogenetic position of ascidians leads to the presumption that neuropeptides and hormones in vertebrates are highly likely to be evolutionarily conserved in ascidians, and the cosmopolitan species Ciona intestinalis is expected to be an excellent deuterostome Invertebrate model
B Schjoldager et al.
The American journal of physiology, 260(6 Pt 1), G976-G982 (1991-06-01)
The protochordean octapeptide cionin is structurally a hybrid of mammalian cholecystokinin (CCK) and gastrin, and thus their possible common ancestor. To determine whether cionin behaves like CCK or gastrin, we examined its effect on canine fundic somatostatin cells and on
R Mårvik et al.
Scandinavian journal of gastroenterology, 29(7), 591-594 (1994-07-01)
The protochordean octapeptide cionin is structurally a hybrid of mammalian cholecystokinin (CCK) and gastrin. An earlier study has shown that cionin in mammals stimulates gallbladder contraction and gastric somatostatin release, similarly to CCK-8. In the present study we examined the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service