Skip to Content
Merck
All Photos(1)

Key Documents

SML2383

Sigma-Aldrich

Isosilybin

≥98% (HPLC)

Synonym(s):

Isosilybin, mixture of isomers, (2R,3R)-2-[2,3-Dihydro-2-(4-hydroxy-3-methoxyphenyl)-3-(hydroxymethyl)-1,4-benzodioxin-6-yl]-2,3-dihydro-3,5,7-trihydroxy-4H-1-benzopyran-4-one, Isosilybinin

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$163.00
25 MG
$524.00

$163.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$163.00
25 MG
$524.00

About This Item

Empirical Formula (Hill Notation):
C25H22O10
CAS Number:
Molecular Weight:
482.44
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$163.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O1C(C(Oc3c1cc(cc3)C4Oc5c(c(cc(c5)O)O)C(=O)C4O)c2cc(c(cc2)O)OC)CO

InChI

1S/C25H22O10/c1-32-17-6-11(2-4-14(17)28)24-20(10-26)33-18-7-12(3-5-16(18)34-24)25-23(31)22(30)21-15(29)8-13(27)9-19(21)35-25/h2-9,20,23-29,31H,10H2,1H3

InChI key

FDQAOULAVFHKBX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Isosilybin is a diastereoisomeric mixture of flavonolignans isosilybin A and isosilybin B isolated from milk thistle (Silybum marianum) that exhibits hepatoprotective activity. Isosilybin potently inhibits Cytochrome P4502C8 (CYP2C8) activity in human liver microsomes. Also Isosilybin is an activator of PPARγ.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eva-Maria Pferschy-Wenzig et al.
Journal of natural products, 77(4), 842-847 (2014-03-07)
Peroxisome proliferator-activated receptor gamma (PPARγ) is a key regulator of glucose and lipid metabolism. Agonists of this nuclear receptor are used in the treatment of type 2 diabetes and are also studied as a potential treatment of other metabolic diseases
Ahmed A Albassam et al.
Chemico-biological interactions, 271, 24-29 (2017-05-02)
Milk thistle is a widely-consumed botanical used for an array of purported health benefits. The primary extract of milk thistle is termed silymarin, a complex mixture that contains a number of structurally-related flavonolignans, the flavonoid, taxifolin, and a number of
David Y-W Lee et al.
Journal of natural products, 66(9), 1171-1174 (2003-09-27)
Two pairs of diastereoisomeric flavonolignans, silybin A, silybin B, isosilybin A, and isosilybin B, were successfully separated from Silybum marianum by sequential silica gel column chromatography, preparative reversed-phase HPLC, and recrystallization. Complete stereochemical assignments at C-2, C-3, C-7', and C-8'

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service