Skip to Content
Merck
All Photos(1)

Key Documents

SCP0099

Sigma-Aldrich

Caspase Substrate

≥95% (HPLC), lyophilized

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$250.00

$250.00


Usually ships in 2 weeks.


Select a Size

Change View
5 MG
$250.00

About This Item

Empirical Formula (Hill Notation):
C31H39N6O10
Molecular Weight:
655.68
UNSPSC Code:
12352202
NACRES:
NA.32

$250.00


Usually ships in 2 weeks.

Product Name

Caspase Substrate,

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥80%

storage condition

protect from light

storage temp.

−20°C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service