Skip to Content
Merck
All Photos(1)

Key Documents

277207

Sigma-Aldrich

Carbon-13C

99 atom % 13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13C
CAS Number:
Molecular Weight:
13.00
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.12
Product 277207 is currently not available for purchase. Contact Technical Service

isotopic purity

99 atom % 13C

Quality Level

form

amorphous

mp

3550 °C (lit.)

mass shift

M+1

SMILES string

[13C]

InChI

1S/C/i1+1

InChI key

OKTJSMMVPCPJKN-OUBTZVSYSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

  1. What is the particle size of this product? Thank you.

    1 answer
    1. Unfortunately, the particle size is not determined for this product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service