Skip to Content
Merck
All Photos(1)

Key Documents

SCP0021

Sigma-Aldrich

Angiotensinogen 1-14 Renin Substrate porcine

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$148.00

$148.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
1 MG
$148.00

About This Item

Empirical Formula (Hill Notation):
C85H123N21O20
Molecular Weight:
1759.01
UNSPSC Code:
12352200
NACRES:
NA.32

$148.00


Usually ships in 2 weeks.

Request a Bulk Order

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥70%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Asp-Arg-Val-Tyr-Ile-His-Pro-Phe-His-Leu-Leu-Val-Tyr-Ser

Application

Renin (angiotensinogenase) which hydrolyzes angiotensinogen is a component of the renin-angiotensin system (RAS) that regulates vasoconstriction and blood pressure. Angiotensinogen 1-14 (DRVYIHPFHLLVYS) may be used as a substrate for renin. A variety of renin substrates may be built around the minimal sequence His-Pro-Phe-His-Leu-Leu-Val-Tyr (HPFHLLVY).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sining Leng et al.
Nature communications, 11(1), 1680-1680 (2020-04-05)
Rosettes are widely used in epithelial morphogenesis during embryonic development and organogenesis. However, their role in postnatal development and adult tissue maintenance remains largely unknown. Here, we show zona glomerulosa cells in the adult adrenal cortex organize into rosettes through

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service