Skip to Content
Merck
All Photos(1)

Key Documents

SCJ4600015

Sigma-Aldrich

CF 488A hydrazide

suitable for fluorescence, ≥90% (HPLC)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$271.00

$271.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
1 MG
$271.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$271.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Quality Level

Assay

≥90% (HPLC)

form

solid

mol wt

~928 g/mol

solubility

H2O: soluble

ε (extinction coefficient)

70000 at 490 nm

absorbance ratio

A280/490 nm 0.1

fluorescence

λex 488 nm (Flow cytometry laser line)
λex 488 nm (Microscopy laser line)
λex 490 nm; λem 515 nm

suitability

suitable for fluorescence

storage temp.

−20°C

General description

CF488A is a green fluorescent dye optimally excitable by the 488 nm argon laser line. Under common detection conditions, CF488A is at least as bright as Alexa Fluor 488. However, a major advantage of CF488A over Alexa Fluor 488 is that antibody conjugates prepared from the former are biologically more specific. Alexa Fluor 488 carries multiple negative charges, which can significantly change the isoelectric point of the proteins the dye labels and consequently alter the specificity of the protein conjugates. CF488A, on the other hand, is minimally charged. Thus, antibody conjugates prepared from the dye ensure biological detection with high signal-to-noise ratio. Another feature of CF488A is that the emission peak wavelength is about 10 nm shorter than that of Alexa Fluor 488 and 15 nm shorter than that of the traditional green dye FITC (or FAM). The shorter wavelength of CF488A offers the advantage of less fluorescence "spill-over" in the red channel in multi-color detection applications.

CF dye hydrazides can be used as a fixable polar tracers for visualizing cell morphology,or for gap junction studies. The dyes can be introduced into cells by microinjection. For conjugation to aldehyde or ketone groups (such as carbohydrate molecules after peroxidation with periodate) we recommend using the more reactive CF aminooxy dye forms.

Features and Benefits

1. Yields biologically more specific antibody conjugates and has less "spill-over" fluorescence in the red channel than Alexa Fluor488. 2. Extremely photostable. 3. Highly water-soluble and pH-insensitive.

Preparation Note

Protect from light

Reconstitution

Dissolve in water

Legal Information

This product is distributed by Sigma-Aldrich Co. under the authorization of Biotium, Inc. This product is covered by one or more US patents and corresponding patent claims outside the US patents or pending applications owned or licensed by Biotium, Inc. including without limitation: 12/334,387; 12/607,915; 12/699,778; 12/850,578; 61/454,484. In consideration of the purchase price paid by the buyer, the buyer is hereby granted a limited, non-exclusive, non-transferable license to use only the purchased amount of the product solely for the buyer′s own internal research in a manner consistent with the accompanying product literature. Except as expressly granted herein, the sale of this product does not grant to or convey upon the buyer any license, expressly, by implication or estoppel, under any patent right or other intellectual property right of Biotium, Inc. Buyer shall not resell or transfer this product to any third party, or use the product for any commercial purposes, including without limitation, any diagnostic, therapeutic or prophylactic uses. This product is for research use only. Any other uses, including diagnostic uses, require a separate license from Biotium, Inc. For information on purchasing a license to use this product for purposes other than research, contact Biotium, Inc., 3159 Corporate Place, Hayward, CA 94545, Tel: (510) 265-1027. Fax: (510) 265-1352. Email: [email protected].
ALEXA FLUOR is a trademark of Life Technologies
CF is a trademark of Biotium, Inc.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service