Skip to Content
Merck
All Photos(1)

Key Documents

SAE0194

Sigma-Aldrich

3xFLAG Peptide

≥95% (HPLC), lyophilized powder, purified

Synonym(s):

Met-Asp-Tyr-Lys-Asp-His-Asp-Gly-Asp-Tyr-Lys-Asp-His-Asp-Ile-Asp-Tyr-Lys-Asp-Asp-Asp-Asp-Lys, ddddk peptide, dykddddk peptide

Sign Into View Organizational & Contract Pricing

Select a Size

4 MG
$604.00

$604.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
4 MG
$604.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.43

$604.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Product Name

Purified 3xFLAG® peptide, ≥95% (HPLC), lyophilized powder

biological source

synthetic

Quality Level

Assay

≥95% (HPLC)

form

lyophilized powder

technique(s)

protein extraction: suitable

shipped in

dry ice

storage temp.

−20°C

General description

The 3X FLAG Peptide is a synthetic peptide of 23 amino acid residues in length. The Asp-Tyr-Lys-Xaa-Xaa-Asp motif is repeated three times in the peptide. Eight amino acids at the C-terminus make up the classic FLAG sequence (Asp-Tyr-Lys-Asp-Asp-Asp-Asp-Lys).

Application

For use in competitive elution by affinity chromatography of 3X FLAG fusion proteins either from the ANTI-FLAG M2 monoclonal antibody (F1804) in solution, or bound to agarose on the ANTI-FLAG M2 agarose affinity gel (A2220).
For elution under gentle, mild conditions of 3X FLAG fusion proteins from the ANTI-FLAG M2 affinity gels.

(Note: the 1X FLAG Peptide (F3290) will not elute 3X FLAG fusion proteins from ANTI-FLAG affinity gels.)

Legal Information

3xFLAG is a trademark of Sigma-Aldrich Co. LLC
FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Find protein research tools to prepare, isolate, and analyze proteins. Organized by how to extract, protect, purify, enrich, modify, and quantify proteins.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service