Skip to Content
Merck
All Photos(1)

Key Documents

SAE0161

Sigma-Aldrich

Albumin Human

Optibumin, Recombinant, Animal Component Free, suitable for cell culture, low lipid

Synonym(s):

Optibumin, rHSA

Sign Into View Organizational & Contract Pricing

Select a Size

10 ML
$258.00

$258.00


Estimated to ship onApril 04, 2025


Request a Bulk Order

Select a Size

Change View
10 ML
$258.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.25

$258.00


Estimated to ship onApril 04, 2025


Request a Bulk Order

biological source

human

Quality Level

mol wt

monomer ~67 kDa

storage temp.

2-8°C

General description

Exbumin is high-purity recombinant human serum albumin with high lysine reactivity. It contains low levels of albumin aggregates and lipids and high concentration of free-thiol/Cys-34 mercapto-albumin. This USA-origin rHSA is animal-free.

Application

Exbumin is the highest purity rHSA available on the market optimized for most demanding applications.


  • solubilization or bioconjugation of molecules
  • cryopreservation of iPSC and hESC

Biochem/physiol Actions

Serum albumin functions as a carrier protein for steroids, fatty acids, and thyroid hormones. They are also vital in regulating the colloidal osmotic pressures of blood. It is added as an ingredient to classical and custom media formulations. Serum albumins protect the cells against physical or toxins-induced stress.

Features and Benefits

Exbumin is animal component free that is manufactured using good manufacturing practices at an ISO9001:2015 certified facility.
Advantages


  • High mercaptoalbumin (Cys34 free thiol)
  • Low lipid content
  • High lysine reactivity
  • Eliminates animal-derived additives
  • Safe and regulatory friendly
  • Provides well-defined and consistent stem cell medium
  • Raw materials, manufacturing equipment, and packaging is certified blood free and animal free

Packaging

10 ml in poly bottle

Other Notes

Exbumin is a registered trademark of InVitria

Legal Information

Exbumin is a trademark of Ventria Bioscience Inc.
Optibumin is a trademark of Ventria Bioscience Inc.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service