Skip to Content
Merck
All Photos(1)

Key Documents

NIST2925

Recombinant Human Serum Albumin Solution (Primary Reference Calibrator for Urine Albumin) (Frozen)

NIST®SRM®

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$1,820.00

$1,820.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$1,820.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$1,820.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

liquid

packaging

pkg of 1 g (2 vials x 0.5mL each)

manufacturer/tradename

NIST®

technique(s)

mass spectrometry (MS): suitable

application(s)

clinical testing

storage temp.

−20°C

General description

Each unit of SRM 2925 consists of two vials, each containing approximately 0.5 mL of HSA in 50 mmol/L ammonium bicarbonate in water. For more information, please refer to the SDS and COA.
SRM 2925_cert
SRM 2925_SDS

Application

Recombinant human serum albumin (HSA) is a standard reference material (SRM) intended for calibration of liquid chromatography-tandem mass spectrometric procedures for the determination of albumin in urine.

Features and Benefits

  • NIST certified value for recombinant HSA was determined through amino acid analysis using (ID-LC-MS/MS) and is expressed as nanomoles per gram, and is metrologically traceable to the SI-derived units for molar concentration.
  • Expiration of Certification, Instructions for handling, storage, and use are provided on the NIST certificate.

Other Notes

  • Details on expiration, storage, safety, usage, and source are provided in the NIST certificate.
  • Notes on biomaterials, disposal, and transport are documented in SDS.
Please download a current certificate at nist.gov/SRM for current analytes and certified values.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jia Wang et al.
STAR protocols, 3(4), 101908-101908 (2023-01-04)
Here, we present a protocol to reprogram mouse and human fibroblasts into expandable cardiovascular progenitor cells (CPCs) via a defined small-molecule treatment. We describe steps to prepare fibroblasts and generate the chemically induced CPCs (ciCPCs), followed by expansion and differentiation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service