Skip to Content
Merck
All Photos(2)

Key Documents

SAE0117

Sigma-Aldrich

Amelogenin, human

X isoform, recombinant, expressed in E. coli

Synonym(s):

AIH1, AMELX, AMG, AMGX, Amel

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.26
Pricing and availability is not currently available.

biological source

human

recombinant

expressed in E. coli

Assay

≥90% (HPLC)

form

solid

impurities

≤1 EU/μg

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... AMELX(265)

General description

Research area: Neuroscience

Amelogenin, X isoform belongs to the family of closely related amelogenin proteins that participate in the amelogenesis process of teeth mineralization. Amelogenins account for more than 90% of the total protein in developing tooth enamel.

Biochem/physiol Actions

Mutations in AMELX are responsible for the development of the rare disease Amelogenesis imperfecta 1E (AI1E), which is characterized by abnormal tooth enamel development. Amelogenins act both as attachment factors and as a growth factor for mesenchymal stem cells. Amelogenin is of increasing importance as a potential therapeutic agent for variety of application including treatment hard-to-heal wounds, promoting remineralization of caries lesions, and repairing human tooth enamel.

Preparation Note

The product can be reconstituted in water or 2% acetic acid in a concentration of 0.1-5 mg/ml. Tap vial to make sure protein is reconstituted. Aliquot reconstituted Amelogenin and store at -20 °C. Avoid repeated freeze/thaw cycles. Amelogenin may form thin clear film on some surfaces, leading to reduced concentration in solution. Exact Amelogenin concentration can be determined by absorption at 280nm. Amelogenin solution of 0.1% has A280=1.35

Other Notes

The product is supplied as lyophilized powder, prepared from a 0.2 μM filtered solution of Amelogenin in 2% acetic acid without any additives. The lyophilized product is essentially salt free. The biological activity of recombinant, human AMELX is measured by its ability to promote attachment of Saos-2 cells to a coated surface. The ED50 is defined as the amount of AMELX per cm2 that elicits 50% of the maximal attachment activity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Lagerström-Fermér et al.
Genomics, 26(1), 159-162 (1995-03-01)
Formation of tooth enamel is a poorly understood biological process. In this study we describe a 9-bp deletion in exon 2 of the amelogenin gene (AMGX) causing X-linked hypoplastic amelogenesis imperfecta, a disease characterized by defective enamel. The mutation results
Regeneration of bone and periodontal ligament induced by recombinant amelogenin after periodontitis.
Haze, A. et al.
Journal of Cellular and Molecular Medicine, 13(6), 1110-1124 (2009)
Maggie Zeichner-David et al.
European journal of oral sciences, 114 Suppl 1, 244-253 (2006-05-06)
Enamel proteins, particularly amelogenin, have been associated with other functions in addition to regulating enamel biomineralization. Extracts of enamel proteins are currently being used to regenerate periodontal tissues, and new studies suggest that enamel proteins might have chondrogenic and osteogenic
Frank Schwarz et al.
Clinical oral investigations, 8(3), 165-171 (2004-04-03)
The purpose of the present study was to investigate the effects of an enamel matrix protein derivative (EMD) on attachment, proliferation, and viability of human SaOs(2) osteoblasts on titanium implants. A total of 220 sand-blasted and acid-etched (SLA) titanium discs
Masanobu Izumikawa et al.
TheScientificWorldJournal, 2012, 879731-879731 (2012-05-02)
The aim of this study was to clarify the function of amelogenin, the major protein of enamel matrix derivative, on the proliferation, differentiation, and mineralization of cultured rat bone marrow stem cells (BMSCs), toward the establishment of future bone regenerative

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service