SAB4700332
Monoclonal Anti-CD54-FITC antibody produced in mouse
clone 1H4, purified immunoglobulin, buffered aqueous solution
Synonym(s):
Anti-ICAM1
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
NACRES:
NA.44
Recommended Products
biological source
mouse
Quality Level
conjugate
FITC conjugate
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
1H4, monoclonal
form
buffered aqueous solution
species reactivity
human
technique(s)
flow cytometry: suitable
isotype
IgG2b
NCBI accession no.
UniProt accession no.
shipped in
wet ice
storage temp.
2-8°C
target post-translational modification
unmodified
Gene Information
human ... ICAM1(3383)
General description
CD54 (cluster of differentiation 54), also known as ICAM1 (intercellular adhesion molecule 1) was initially identified as a ligand for the β2 integrin lymphocyte function-associated antigen (LFA)-1. It is a glycoprotein with a 90,000-110,000 molecular weight and shows wide range of tissue expression, including tissue macrophages and dendritic cells. It also shows expression on non-hematopoietic cells such as vascular endothelial cells, epithelial cells of thymus and mucosa and dermal fibroblasts. It is a member of the ICAM family, which also includes ICAM-2, -3, -4 and -5.
The antibody 1H4 reacts with CD54 (ICAM-1), a 85-110 kDa type I transmembrane glycoprotein (receptor for rhinovirus) expressed on activated endothelial cells, T lymphocytes, B lymphocytes, monocytes, macrophages, granulocytes and dendritic cells; the expression of CD54 is upregulated by activation.
Immunogen
Raji cells and spleen cells fused with NS1 cells
Application
The reagent is designed for Flow Cytometry analysis of human blood cells using 20 μL reagent / 100 μL of whole blood or 1e6 cells in a suspension. The content of a vial (2 mL) is sufficient for 100 tests.
Biochem/physiol Actions
The expression of CD54 (cluster of differentiation 54)/ICAM1 (intercellular adhesion molecule 1) is strongly increased by interleukin-1 (IL-1) and interferon (IFN)-γ. It is expressed on lymphoid cells where it is required for phorbol ester-induced aggregation. It participates in lymphocyte function-associated antigen 1 (LFA-1)-dependent NK cell stimulation through its attachment to actin cytoskeleton through α-actinin or ezrin. It also acts as the receptor for rhinovirus, a common cold virus and Plasmodium falciparum. Soluble ICAM-1 levels are elevated in pregnant women with gestational diabetes mellitus (GDM), which is an outcome of endothelial cell activation and dysfunction. Meta-analysis of case-control studies show that rs1799969 polymorphism in ICAM-1 gene is linked with increased susceptibility to cancer.
Features and Benefits
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.
Physical form
Solution in phosphate buffered saline containing 15 mM sodium azide and 0.2% high-grade protease free BSA as a stabilizing agent.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
ICAM-1: getting a grip on leukocyte adhesion.
Long EO
Journal of Immunology, 186, 5021-5023 (2011)
ICAM-1 (CD54): a counter-receptor for Mac-1 (CD11b/CD18).
Diamond MS
The Journal of Cell Biology, 111, 3129-3139 (1990)
Intercellular Adhesion Molecule and Endogenous NOS Inhibitor: Asymmetric Dimethylarginine in Pregnant Women with Gestational Diabetes Mellitus.
Poniedzialek-Czajkowska E
Journal of Diabetes Research (2016)
Association of Polymorphisms in Intercellular Adhesion Molecule 1 (ICAM-1) Gene with Cancer Susceptibility: A Meta-Analysis of 14 Case-Control Studies.
Zhang X et al
Medical Science Monitor : International Medical Journal of Experimental and Clinical Research, 22, 569-579 (2016)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service