Skip to Content
Merck
All Photos(1)

Key Documents

FCMAB198F

Sigma-Aldrich

Milli-Mark® Anti-CD54-FITC Antibody, clone 6.5B5

clone 6.5B5, Milli-Mark®, from mouse

Synonym(s):

Intercellular adhesion molecule 1, ICAM-1, Major group rhinovirus receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.44
Pricing and availability is not currently available.

biological source

mouse

Quality Level

conjugate

FITC conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6.5B5, monoclonal

species reactivity

human

manufacturer/tradename

Milli-Mark®

technique(s)

flow cytometry: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ICAM1(3383)

General description

The CD54 antigen is a single chain glycoprotein with a molecular mass of 90 kDa (Springer, 1990). It is the human intercellular adhesion molecule-1 (ICAM-1) belonging to the immunoglobulin supergene family. The ligand for ICAM-1 is LFA-1 (leucocyte function associated-1 protein) (Berendt, 1992). ICAM-1 is widely expressed on many cell types, although the degree of expression by unstimulated resting cells varies. As a cellular activation antigen the expression of ICAM-1 is up-regulated upon cell activation, being induced particularly in response to cytokines such as interleukin-1, tumor necrosis factor, and interferon- (Berendt, 1992; Wellicome, 1990; Thornhill, 1990). ICAM-1 might be a receptor for rhinovirus (Staunton, 1989). In rejecting kidneys the antibody stains all infiltrating cells strongly as well as glomerulus epithelium, endothelium on capillaries, large vessels, and mesangium (Waugh, 1989).

Specificity

Antibody recognizes Human CD54 antigen.

Immunogen

TNF-activated human umbilical vein endothelial cells

Application

Milli-Mark Anti-CD54-FITC Antibody, clone 6.5B5 is an antibody against CD54 for use in FC.
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology

Quality

Evaluated by flow cytometry using human lymphocytes

Target description

90 kDa (Springer, 1990)

Physical form

Protein A purified
Purified Mouse monoclonal IgG1 conjugated to FITC in 0.05M Tris-HCl buffer with 1% BSA, Sodium Azide at 0.097% at pH 7.2.

Storage and Stability

Maintain refrigerated at 2-8°C protected from light in undiluted aliquots for up to 6 months from date of receipt.

Analysis Note

Control
Human Lymphocytes

Legal Information

MILLI-MARK is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service