Skip to Content
Merck
All Photos(2)

Key Documents

SAB4503802

Sigma-Aldrich

Anti-phospho-BTK (pTyr551) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-AGMX1, Anti-AT, Anti-ATK, Anti-BPK, Anti-IGHD3, Anti-IMD1, Anti-PSCTK1, Anti-XLA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 76 kDa

species reactivity

rat, human, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:40000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr551)

Gene Information

human ... BTK(695)

General description

Bruton tyrosine kinase (BTK) is encoded by the gene mapped to human chromosome Xq22. The encoded protein belongs to the Tec family of kinases.

Immunogen

The antiserum was produced against synthesized peptide derived from human BTK around the phosphorylation site of Tyr551.

Immunogen Range: 516-565

Biochem/physiol Actions

Bruton tyrosine kinase (BTK) is a key element of B-cell–receptor signaling, which facilitates interactions with the tumor microenvironment and stimulates the survival and proliferation of chronic lymphocytic leukemia (CLL) cells. The encoded protein also plays a crucial role in activation of various constitutively active pathways of CLL-cell survival, such as the Akt, extracellular signal-regulated kinase (ERK) and nuclear factor κ light-chain enhancer of activated B cells (NF-κB) pathways. Additionally, it is also necessary for chemokine-mediated homing and adhesion of B cells. Polymorphisms in the gene are associated with the development of a symptomatic primary antibody deficiency (PAD), X-linked agammaglobulinemia.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Targeting BTK with Ibrutinib in Relapsed Chronic Lymphocytic Leukemia
Byrd JC
The New England Journal of Medicine, 369, 32-42 (2013)
The lack of BTK does not impair monocytes and polymorphonuclear cells functions in X-linked agammaglobulinemia under treatment with intravenous immunoglobulin replacement
Cavaliere FM
PLoS ONE, 12 (2017)
Validation of the STR DXS7424 and the linkage situation on the X-chromosome.
Edelmann J
Forensic Science International, 125, 217-222 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service