MAB2223
Anti-DNP Antibody, clone 9H8.1
clone 9H8.1, from mouse
Synonym(s):
Dinitrophenol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Recommended Products
biological source
mouse
Quality Level
antibody form
purified immunoglobulin
clone
9H8.1, monoclonal
species reactivity (predicted by homology)
all
technique(s)
western blot: suitable
isotype
IgG1κ
shipped in
wet ice
target post-translational modification
unmodified
General description
Dinitrophenol (DNP) is a small water-soluble, hydrophobic ligand that binds to transthyretin at acidic pH and also reduces the formation of amyloid fibril. DNP levels of 100 mg/l or more is greatly toxic to acclimated bacteria. DNP has been associated with possible therapies for neurodegenerative and neurological disorders.
Specificity
Demonstrated to react with a DNP moiety in a wider range of species. This antibody′s reactivity is expected to be species independent.
Immunogen
KLH-conjugated DNP
Application
This Anti-DNP Antibody, clone 9H8.1 is validated for use in WB for the detection of DNP.
Quality
Evaluated by Western Blot in Oxyblot protein standards.
Western Blot Analysis: 0.5 µg/mL of this antibody detected DNP on 10 µg of Oxyblot protein standards.
Western Blot Analysis: 0.5 µg/mL of this antibody detected DNP on 10 µg of Oxyblot protein standards.
Target description
Various kDa observed. The Oxyblot protein standards (S7151) were used to test this antibody. These standards are phosphorylase B, BSA, OVA, Carbonic Anhydrase, and trypsin inhibitor which all contain 1-3 DNP residues per protein.
Physical form
Format: Purified
Analysis Note
Control
Oxyblot protein standards
Oxyblot protein standards
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Jennifer B M Steffen et al.
The Journal of experimental biology, 226(15) (2023-07-20)
Coastal environments commonly experience fluctuations in salinity and hypoxia-reoxygenation (H/R) stress that can negatively affect mitochondrial functions of marine organisms. Although intertidal bivalves are adapted to these conditions, the mechanisms that sustain mitochondrial integrity and function are not well understood.
Linda Adzigbli et al.
Scientific reports, 14(1), 9658-9658 (2024-04-27)
Sessile benthic organisms like oysters inhabit the intertidal zone, subject to alternating hypoxia and reoxygenation (H/R) episodes during tidal movements, impacting respiratory chain activities and metabolome compositions. We investigated the effects of constant severe hypoxia (90 min at ~ 0% O2 ) followed
Tara A Seibert et al.
Scientific reports, 14(1), 1029-1029 (2024-01-11)
We evaluated biochemical changes in skeletal muscle of women with breast cancer initiating aromatase inhibitors (AI), including oxidation of ryanodine receptor RyR1 and loss of stabilizing protein calstabin1, and detailed measures of muscle function. Fifteen postmenopausal women with stage I-III breast
Ana Reis-Mendes et al.
Pharmaceuticals (Basel, Switzerland), 14(6) (2021-06-03)
Mitoxantrone (MTX) is a pharmaceutical drug used in the treatment of several cancers and refractory multiple sclerosis (MS). Despite its therapeutic value, adverse effects may be severe, namely the frequently reported cardiotoxicity, whose mechanisms need further research. This work aimed
Rita Araújo et al.
Bone, 158, 116355-116355 (2022-02-14)
Diabetes mellitus (DM) embrace a group of chronic metabolic conditions with a high morbidity, causing deleterious effects in different tissues and organs, including bone. Hyperglycemia seems to be one of the most contributing etiological factors of bone-related alterations, altering metabolic
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service