SAB4502117
Anti-RXFP3 antibody produced in rabbit
affinity isolated antibody
Synonym(s):
G protein-coupled receptor SALPR, GPCR135, RLN3 receptor 1, Relaxin family peptide receptor 3, Somatostatin- and angiotensin-like peptide receptor
About This Item
Recommended Products
biological source
rabbit
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
mol wt
antigen 51 kDa
species reactivity
rat, human, mouse
concentration
~1 mg/mL
technique(s)
ELISA: 1:10000
western blot: 1:500-1:1000
NCBI accession no.
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... RXFP3(51289)
General description
RXFP3 (relaxin family peptide receptor 3) is also termed as GPCR 135/ SALPR (somatostatin- and angiotensin- like peptide receptor). It is distributed in the central nervous system (CNS), mainly in areas associated with memory, sensory and emotional processing. RXFP3 belongs to the insulin superfamily. This gene is mapped to human chromosome 5p13.
Immunogen
Immunogen Range: 161-210
Biochem/physiol Actions
Features and Benefits
Physical form
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service