Merck
Relaxin-3/RXFP3 system regulates alcohol-seeking.
Proceedings of the National Academy of Sciences of the United States of America (2013-12-04)
Philip J Ryan, Hanna E Kastman, Elena V Krstew, K Johan Rosengren, Mohammed Akhter Hossain, Leonid Churilov, John D Wade, Andrew L Gundlach, Andrew J Lawrence
ABSTRACT
Relapse and hazardous drinking represent the most difficult clinical problems in treating patients with alcohol use disorders. Using a rat model of alcohol use and alcohol-seeking, we demonstrated that central administration of peptide antagonists for relaxin family peptide 3 receptor (RXFP3), the cognate receptor for the highly conserved neuropeptide, relaxin-3, decreased self-administration of alcohol in a dose-related manner and attenuated cue- and stress-induced reinstatement following extinction. By comparison, RXFP3 antagonist treatment did not significantly attenuate self-administration or reinstatement of sucrose-seeking, suggesting a selective effect for alcohol. RXFP3 is densely expressed in the stress-responsive bed nucleus of the stria terminalis, and bilateral injections of RXFP3 antagonist into the bed nucleus of the stria terminalis significantly decreased self-administration and stress-induced reinstatement of alcohol, suggesting that this brain region may, at least in part, mediate the effects of RXFP3 antagonism. RXFP3 antagonist treatment had no effect on general ingestive behavior, activity, or procedural memory for lever pressing in the paradigms assessed. These data suggest that relaxin-3/RXFP3 signaling regulates alcohol intake and relapse-like behavior, adding to current knowledge of the brain chemistry of reward-seeking.
Product Description
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Millipore
Sucrose, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, Grade I, ≥99% (GC), suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)