Skip to Content
Merck
All Photos(1)

Key Documents

SAB4501401

Sigma-Aldrich

Anti-HMGB1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

HMG-1, HMG1, High mobility group protein 1, High mobility group protein B1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 24 kDa

species reactivity

rat, human, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HMGB1(3146)

General description

Anti-HMGB1 Antibody detects endogenous levels of total HMGB1 protein.
High mobility group box 1 (HMGB1) is a nuclear protein that belongs to the damage-associated molecular pattern (DAMP) family. It is located on human chromosome 13q12.

Immunogen

The antiserum was produced against synthesized peptide derived from human HMGB1.

Immunogen Range: 131-180

Biochem/physiol Actions

High mobility group box 1 (HMGB1) is involved in bone remodeling and ectopic calcification pathogenesis. It initiates neuroinflammation after epileptogenic injuries. HMGB1 also binds to chromatin and modulates gene transcription.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecular isoforms of high-mobility group box 1 are mechanistic biomarkers for epilepsy.
Walker LE, et al.
The Journal of Clinical Investigation, 127(6), 2118-2132 (2017)
Roles of High Mobility Group Box 1 in Cardiovascular Calcification.
Chen Q, et al.
Cellular Physiology and Biochemistry, 42(2), 427-440 (2017)
A new case of 13q12.2q13.1 microdeletion syndrome contributes to phenotype delineation.
Mandrile G,et al.
Case Reports in Genetics (2014)
Laura Patricia Whittall-Garcia et al.
Lupus science & medicine, 11(1) (2024-01-05)
To determine if the serum levels of neutrophil extracellular trap (NET) remnants (Elastase-DNA and HMGB1-DNA complexes) at the time of a lupus nephritis (LN) flare predict renal outcomes in the following 24 months. This was a retrospective study performed in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service