Skip to Content
Merck
All Photos(2)

Documents

A5533

Sigma-Aldrich

Alizarin Red S

certified by the Biological Stain Commission, powder

Synonym(s):

3,4-Dihydroxy-9,10-dioxo-2-anthracenesulfonic acid sodium salt, Alizarin Carmine, Alizarin Red, water soluble, Alizarin sodium monosulfonate, Alizarin sulfonate sodium, Alizarinsulfonic acid sodium salt, Sodium alizarinsulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H7NaO7S
CAS Number:
Molecular Weight:
342.26
Colour Index Number:
58005
Beilstein:
4117090
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

product name

Alizarin Red S, certified by the Biological Stain Commission

grade

certified by the Biological Stain Commission

Quality Level

form

powder

concentration

0.015 g/L in 0.1 M NaOH

technique(s)

titration: suitable

color

dark red
yellow-orange to dark orange

solubility

H2O: 1 mg/mL

ε (extinction coefficient)

592-598 nm at 0.015 g/L in 0.1 M NaOH

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].Oc1c(O)c(cc2C(=O)c3ccccc3C(=O)c12)S([O-])(=O)=O

InChI

1S/C14H8O7S.Na/c15-11-6-3-1-2-4-7(6)12(16)10-8(11)5-9(22(19,20)21)13(17)14(10)18;/h1-5,17-18H,(H,19,20,21);/q;+1/p-1

InChI key

HFVAFDPGUJEFBQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

  • Alizarin Red S has been used for checking the calcium deposits in osteogenic culture.
  • It has been used for the quantitative analyses of calcification in aortic explants.
  • It has been used for mineral staining in osteoblast cultures.

Biochem/physiol Actions

Alizarin Red S is used for the staining of cartilage and bone. It reacts with calcium, thereby helping in the diagnosis of calcium deposits. It is also used to study carbohydrate–boronic acid interactions.

Suitability

Certified for use in Dahl′s method for calcium

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Impaired marrow osteogenesis is associated with reduced endocortical bone formation but does not impair periosteal bone formation in long bones of SAMP6 mice.
Silva MJ, et al.
Journal of Bone and Mineral Research, 20, 419-419 (2005)
Pathologic calcification of adult vascular smooth muscle cells differs on their crest or mesodermal embryonic origin.
Leroux-Berger M, et al.
Journal of Bone and Mineral Research, 26, 1543-1543 (2011)
RGD peptides immobilized on a mechanically deformable surface promote osteoblast differentiation.
Cavalcanti-Adam EA, et al.
Journal of Bone and Mineral Research, 17, 2130-2130 (2002)
On the history and mechanism of alizarin and alizarin red S stains for calcium.
Puchtler H, et al.
The Journal of Histochemistry and Cytochemistry, 17, 110-124 (1969)
Phenol red inhibits chondrogenic differentiation and affects osteogenic differentiation of human mesenchymal stem cells in vitro.
Lysdahl H, et al.
Stem Cell Reviews, 9, 132-132 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service