Skip to Content
Merck
All Photos(3)

Key Documents

SAB4200641

Sigma-Aldrich

Monoclonal Anti-Brg1 antibody produced in rat

clone 3G4, purified from hybridoma cell culture

Synonym(s):

Anti-ATP-dependent helicase SMARCA4, Anti-BAF190, Anti-BAF190A, Anti-BRG1, Anti-BRG1-associated factor 190A, Anti-BRM/SWI2-related gene 1, Anti-MRD16, Anti-RTPS2, Anti-SMARCA4, Anti-SNF2, Anti-SNF2-beta, Anti-SNF2-like 4, Anti-SNF2L4, Anti-SNF2LB, Anti-SWI/SNF related, Anti-SWI2, Anti-actin dependent regulator of chromatin, Anti-brahma protein-like 1, Anti-global transcription activator homologous sequence, Anti-hSNF2b, Anti-homeotic gene regulator, Anti-matrix associated, Anti-member 4, Anti-mitotic growth and transcription activator, Anti-nuclear protein GRB1, Anti-protein BRG-1, Anti-protein brahma homolog 1, Anti-subfamily a, Anti-sucrose nonfermenting-like 4, Anti-transcription activator BRG1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3G4, monoclonal

form

buffered aqueous solution

mol wt

~185 kDa

species reactivity

mouse, human

concentration

~1 mg/mL

technique(s)

flow cytometry: 2.5-5 μg/test using HeLa cells
immunoblotting: 0.1-0.2.0 μg/mL using HeLa nuclear extract
immunofluorescence: 10 μg/mL using HeLa cells

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BRG1(6597)
mouse ... Brg1(20586)

General description

Monoclonal Anti-Brg1 (rat IgG1 isotype) is derived from the hybridoma 3G4 produced by the fusion of mouse myeloma cells (SP2) and lymph node cells from rat immunized with a synthetic peptide corresponding to a sequence at the C-terminal region of human Brm-related gene-1 (Brg1) protein. Brg1 or SWI/SNF (SWItch/Sucrose non-fermentable) related, matrix-associated, actin-dependent, regulator of chromatin, subfamily A, member 4 (SMARCA4) gene with 35 exons, is encoded by the gene mapped to human chromosome 19p13.

Immunogen

a synthetic peptide corresponding to a sequence at the C-terminal region of human Brg1 protein

Application

Monoclonal Anti-Brg1 antibody produced in rat has been used in:
  • immunoblotting (~185 kDa)
  • flow cytometry
  • immunofluorescence

Biochem/physiol Actions

Brm-related gene-1 (Brg1) (or hBrm) protein is the central catalytic ATPase of the SWI/SNF chromatin-remodeling complex, which is involved in transcriptional modulation of hormone-responsive promoters through binding of the complex to various nuclear receptors and its recruitment to gene-specific promoters. In addition, BRG1 has also been shown to act as a transcriptional repressor. It interacts with retinoblastoma tumor suppressor to form a repressor complex which inhibits cell cycle proteins such as cyclins A, D1 and E. BRG1 participates in the firing of origins and the process of replication elongation. This function of BRG1 also facilitates embryogenesis. In addition, It functions as a tumor suppressor and maintains genome stability and prevent cancer.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The BRG1 transcriptional coregulator
Trotter KW and Archer TK
Nuclear Receptor Signaling, 6(1), nrs-06004 (2008)
Mutation analysis of the BRG1 gene in prostate cancer clinical samples
Valdman A, et al.
International Journal of Oncology, 22(5), 1003-1007 (2003)
Stephanie M Cohen et al.
Nucleic acids research, 38(20), 6906-6919 (2010-06-24)
For DNA replication to occur, chromatin must be remodeled. Yet, we know very little about which proteins alter nucleosome occupancy at origins and replication forks and for what aspects of replication they are required. Here, we demonstrate that the BRG1
H S Zhang et al.
Cell, 101(1), 79-89 (2000-04-25)
We present evidence that Rb forms a repressor containing histone deacetylase (HDAC) and the hSWI/SNF nucleosome remodeling complex, which inhibits transcription of genes for cyclins E and A and arrests cells in the G1 phase of the cell cycle. Phosphorylation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service