Merck
  • Exit from G1 and S phase of the cell cycle is regulated by repressor complexes containing HDAC-Rb-hSWI/SNF and Rb-hSWI/SNF.
Exit from G1 and S phase of the cell cycle is regulated by repressor complexes containing HDAC-Rb-hSWI/SNF and Rb-hSWI/SNF.
Cell (2000-04-25)
H S Zhang, M Gavin, A Dahiya, A A Postigo, D Ma, R X Luo, J W Harbour, D C Dean
ABSTRACT
We present evidence that Rb forms a repressor containing histone deacetylase (HDAC) and the hSWI/SNF nucleosome remodeling complex, which inhibits transcription of genes for cyclins E and A and arrests cells in the G1 phase of the cell cycle. Phosphorylation of Rb by cyclin D/cdk4 disrupts association with HDAC, relieving repression of the cyclin E gene and G1 arrest. However, the Rb-hSWI/SNF complex persists and is sufficient to maintain repression of the cyclin A and cdc2 genes, inhibiting exit from S phase. HDAC-Rb-hSWI/SNF and Rb-hSWI/SNF then appear to maintain the order of cyclin E and A expression during the cell cycle, which in turn regulates exit from G1 and from S phase, respectively.