Skip to Content
Merck
All Photos(1)

Key Documents

SAB3501278

Sigma-Aldrich

Anti-Puumala Virus Nucleocapsid antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$526.00

$526.00


Estimated to ship onApril 19, 2025



Select a Size

Change View
100 μG
$526.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$526.00


Estimated to ship onApril 19, 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

species reactivity

virus (Puumala virus)

concentration

1 mg/mL

technique(s)

ELISA: suitable

isotype

IgG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Puumala virus ... PUUVsSgp1(2943083)

General description

Puumala virus (PUUV) is a rodent-borne hantavirus of the family Bunyaviridae, an enveloped, negative-sense RNA viruses with a tripartite genome that can cause hantavirus pulmonary syndrome (HPS) and is highly homologous to the protype hantavirus Hantaan virus. Like other hantaviruses, the PUUV glycoprotein is synthesized as a precursor that is posttranslationally processed into two glycoproteins G1 (Gn) and G2 (Gc). These glycoproteins interact with the PUUV nucleocapsid (NP) protein through their cytoplasmic tail, and this association has been suggested to be crucial to the binding of the ribonucleoprotein of the PUUV and the assembly of the virus particle.

Immunogen

Puumala virus nucleocapsid antibody was raised against a 19 amino acid peptide near the amino terminus of the Puumala virus nucleocapsid.The immunogen is located within amino acids 130 - 180 of Puumala Virus Nucleocapsid.

Application

Puumala virus nucleocapsid antibody can detect 10ng Puumala virus nucleocapsid peptide in ELISA at 1 μg/ml.

Physical form

Supplied in PBS containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service