Skip to Content
Merck
All Photos(1)

Key Documents

SAB3501276

Sigma-Aldrich

Anti-Hantaan Virus Nucleocapsid antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$526.00

$526.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
100 μG
$526.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

$526.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

species reactivity

virus (Hantaan virus)

concentration

1 mg/mL

technique(s)

ELISA: suitable

isotype

IgG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Hantaan virus ... HTNVsSgp1(2943078)

General description

Hantaan virus (HNTV) is the prototype virus of the genus hantavirus of the family Bunyaviridae, an enveloped, negative-sense RNA virus that is the eiological agent of Korean hemorrhagic fever. Infection of cultured Vero E6 cells with HNTV induced expression of the heat shock protein HSP70. A substantial fraction of HSP70 was found to be associated with the HNTV nucleocapsid (NP). Vero E6 cells that overexpress HSP70 show significantly reduced levels of NP and an increased resistance to infection by HNTV.

Immunogen

Hantaan virus nucleocapsid antibody was raised against an 18 amino acid peptide near the amino terminus of the Hantaan virus nucleocapsid.The immunogen is located within the first 50 amino acids of Hantaan Virus Nucleocapsid.

Application

Hantaan virus nucleocapsid antibody can detect 10ng Hantaan virus nucleocapsid peptide in ELISA at 1 μg/ml.

Physical form

Supplied in PBS containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service