Skip to Content
Merck
All Photos(1)

Key Documents

SAB3501266

Sigma-Aldrich

Anti-Heartland Virus Glycoprotein 1 antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$526.00

$526.00


Estimated to ship onApril 11, 2025



Select a Size

Change View
100 μG
$526.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

$526.00


Estimated to ship onApril 11, 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

species reactivity

virus (Heartland virus)

concentration

1 mg/mL

technique(s)

ELISA: suitable

isotype

IgG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Heartland Virus Glycoprotein 1 Antibody: The Heartland virus is a newly identified member of the phlebovirus genus of the Bunyaviridae family. Initial reports indicate that symptoms may include fever, fatigue, diarrhea, thrombocytopenia, and leukopenia. The Bunyaviridae family of viruses are spherical enveloped viruses with a tripartite RNA genome of negative or ambisense polarity. The glycoprotein RNA encodes a polyprotein that is cleaved into the G1 and G2 proteins.

Immunogen

Heartland virus glycoprotein 1 antibody was raised against a 19 amino acid peptide near the carboxy terminus of the Heartland virus glycoprotein 1.The immunogen is located within amino acids 520 - 570 of Heartland Virus Glycoprotein 1.

Application

Heartland virus glycoprotein 1 antibody can detect 10 ng Heartland virus glycoprotein 1 in ELISA at 1 μg/mL.

Physical form

Supplied in PBS containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service