Skip to Content
Merck
All Photos(1)

Key Documents

SAB3501174

Sigma-Aldrich

Monoclonal Anti-HIV-1 p24 [7F4] (HRP) antibody produced in mouse

purified antibody (Protein A)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$526.00

$526.00


Estimated to ship onMay 24, 2025



Select a Size

Change View
100 μG
$526.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$526.00


Estimated to ship onMay 24, 2025


biological source

mouse

Quality Level

conjugate

horse radish peroxidase (HRP) conjugate

antibody form

purified antibody (Protein A)

antibody product type

primary antibodies

clone

monoclonal

form

liquid

species reactivity

human immunodeficiency virus 1

concentration

1 mg/mL

technique(s)

ELISA: suitable
flow cytometry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human immunodeficiency virus 1 ... GAG(155030)

General description

HIV-1 p24 Monoclonal Antibody: The human immunodeficiency virus type 1 (HIV-1) particle consists of an envelope, a core and the region between the two termed matrix. The HIV-1 Gag protein is a late structural protein that contains four proteins: matrix (p17), capsid (p24), nucleocapsid (p7) and the p6 protein. The p24 constitutes the major core component of the virus and shows high degree of sequence conservation among HIV isolates. The Gag p24 has been used as an integral part of multicomponent HIV-1 vaccines.

Immunogen

Mouse monoclonal HIV-1 p24 antibody was raised against a recombinant full-length HIV-1 p24 protein.

Application

The antibody can be used for detection of p24 by Western blot or ELISA at 0.2 – 0.5μg/mL.

Physical form

Supplied in PBS containing 1% BSA and 0.02% thimerosol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service