Skip to Content
Merck
All Photos(2)

Key Documents

700080P

Avanti

7α,25-dihydroxycholesterol

Avanti Research - A Croda Brand

Synonym(s):

cholest-5-ene-3β,7α,25-triol

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$405.00

$405.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$405.00

About This Item

Empirical Formula (Hill Notation):
C27H46O3
CAS Number:
Molecular Weight:
418.65
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$405.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 1 mg (700080P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@H]1[C@H]2[C@H]3[C@@]([C@H](CC3)[C@@H](CCCC(O)(C)C)C)(CC[C@@H]2[C@]4(CC[C@@H](CC4=C1)O)C)C

InChI

1S/C27H46O3/c1-17(7-6-12-25(2,3)30)20-8-9-21-24-22(11-14-27(20,21)5)26(4)13-10-19(28)15-18(26)16-23(24)29/h16-17,19-24,28-30H,6-15H2,1-5H3/t17-,19+,20-,21+,22+,23-,24+,26+,27-/m1/s1

InChI key

BQMSKLCEWBSPPY-IKVTXIKFSA-N

General description

7α,25-dihydroxycholesterol (7α,25-OHC) is synthesized by the hydroxylation of cholesterol by the action of enzyme cholesterol 25-hydroxylase (CH25H) and cytochrome P450, family 7, subfamily b, polypeptide 1 (CYP7B1).[1] The catabolic breakdown of 7α,25-OHC to bile acid precursors is catalyzed by the enzymes hydroxy-Δ-5-steroid dehydrogenase, 3β- and steroid Δ-isomerase 7 (HSD3B7).[2]

Application

7α,25-dihydroxycholesterol has been used:
  • as a G-protein-coupled receptor 183 ( GPR183) ligand in chemotaxis assay and intracellular cytokine staining method[2] and in fluorescence-activated cell sorting (FACS) analysis of natural killer cells[3]
  • in oxysterol based calcium mobilization assay in Chinese hamster ovary (CHO) cells[1]
  • in competitive radioligand binding assay of Epstein-Barr virus-induced gene 2 (EBI2) expressing COS-7 cells[4]

Biochem/physiol Actions

7α,25-dihydroxycholesterol (7α,25-OHC) serves as an endogenous ligand for G-protein-coupled receptor 183 (GPR183) or Epstein-Barr virus-induced gene 2 (EBI2). It plays a key role in the migration of type 3 innate lymphoid cells (ILC3) in the small intestine and colon. 7α,25-OHC mediates immune cell migration functionality by their chemoattractant property.[1] Inhibition of 7α,25-OHC synthesis leads to impairment in the migration of B cells.[4]

Packaging

5 mL Amber Glass Screw Cap Vial (700080P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gene regulatory programs conferring phenotypic identities to human NK cells
Collins PL, et al.
Cell, 176(1-2), 348-360 (2019)
Identification of structural motifs critical for epstein-barr virus-induced molecule 2 function and homology modeling of the ligand docking site
Zhang L, et al.
Molecular Pharmacology, 82(6), 1094-1103 (2012)
Oxysterol sensing through the receptor GPR183 promotes the lymphoid-tissue-inducing function of innate lymphoid cells and colonic inflammation
Emgaard J, et al.
Immunity, 48(1), 120-132 (2018)
Katharina R Beck et al.
The Journal of steroid biochemistry and molecular biology, 190, 19-28 (2019-03-25)
Oxysterols are cholesterol metabolites derived through either autoxidation or enzymatic processes. They consist of a large family of bioactive lipids that have been associated with the progression of multiple pathologies. In order to unravel (patho-)physiological mechanisms involving oxysterols, it is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service