Skip to Content
Merck
All Photos(3)

Key Documents

SAB3500126

Sigma-Aldrich

Monoclonal Anti-ORAI1 antibody produced in mouse

clone 3F6H5, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-Calcium release-activated calcium channel protein 1, Anti-TMEM142A, Anti-Transmembrane protein 142A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3F6H5, monoclonal

form

buffered aqueous solution

mol wt

predicted mol wt 33 kDa

species reactivity

human, rat, mouse

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ORAI1(84876)

Immunogen

raised against a 16 amino acid peptide from near the carboxy terminus of human ORAI1.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Antigen stimulation of immune cells triggers Ca++ entry through Ca++ release-activated Ca++ (CRAC) channels. ORAI1 is a recently identified four-transmembrane spanning protein that is an essential component of CRAC. A missense mutation in this protein in humans is the cause of one fo rm of hereditary severe combined immune deficiency (SCID) which results in ablated T-cell Ca++ entry. It has been suggested that ORAI1 functions as a highly selective Ca++ plasma membrane channel that is gated through interactions with STIM1, the store-activated endoplasmic reticulum Ca++ sensor. ORAI1 often migrates at a higher than expected molecular weight in SDS-PAGE. This antibody is predicted to have no cross-reactivity to ORAI2 or ORAI3.

Physical form

Supplied at approx. 1 mg/mL in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Momin Mohis et al.
American journal of physiology. Heart and circulatory physiology, 315(1), H83-H91 (2018-07-10)
Senescence-related fibrosis contributes to cardiac dysfunction. Profibrotic processes are Ca2+ dependent. The effect of aging on the Ca2+ mobilization processes of human ventricular fibroblasts (hVFs) is unclear. Therefore, we tested whether aging altered intracellular Ca2+ release and store-operated Ca2+ entry
Agnese Secondo et al.
Stroke, 50(5), 1240-1249 (2019-04-23)
Background and Purpose- Disturbance of endoplasmic reticulum (ER) Ca2+ homeostasis causes neuronal cell injury in stroke. By contrast, ischemic preconditioning (IPC)-a brief sublethal ischemic episode affording tolerance to a subsequent ischemic insult-restores ER Ca2+ homeostasis. Under physiological conditions, ER calcium
Guangyu Cheng et al.
Frontiers in molecular neuroscience, 17, 1391189-1391189 (2024-07-04)
This investigation aims to elucidate the novel role of Stromal Interaction Molecule 1 (STIM1) in modulating store-operated calcium entry (SOCE) and its subsequent impact on inflammatory cytokine release in T lymphocytes, thereby advancing our understanding of trigeminal neuralgia (TN) pathogenesis.
Gracious R Ross et al.
Biology open, 6(3), 326-332 (2017-01-28)
Excessive cardiac fibrosis, characterized by increased collagen-rich extracellular matrix (ECM) deposition, is a major predisposing factor for mechanical and electrical dysfunction in heart failure (HF). The human ventricular fibroblast (hVF) remodeling mechanisms that cause excessive collagen deposition in HF are
Ana Paula Arruda et al.
eLife, 6 (2017-12-16)
Defective Ca2+ handling is a key mechanism underlying hepatic endoplasmic reticulum (ER) dysfunction in obesity. ER Ca2+ level is in part monitored by the store-operated Ca2+ entry (SOCE) system, an adaptive mechanism that senses ER luminal Ca2+ concentrations through the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service