Skip to Content
Merck
All Photos(1)

Key Documents

A-VMPAKMP

Sigma-Aldrich

Universal Coupling Kit

Sign Into View Organizational & Contract Pricing

Select a Size

50 REACTIONS
$707.00

$707.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 REACTIONS
$707.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.22

$707.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

usage

sufficient for 50 reactions

availability

not available in Japan

storage temp.

2-8°C

General description

The Universal Coupling Kit makes particle-based immunoassays, lateral flow tests and biomolecule separation applications more flexible than ever before. It is the only kit that allows users to select and couple their choice of carboxylated particle with their chosen protein.


Employing a unique mechanism to immobilise proteins, Anteo′s advantages outweigh those of conventional covalent chemistries such as NHS/EDC or passive binding. This facilitates coupling of antibodies with ease, improved functionality and reproducibility, leading to better uniformity between experiments.


Anteo′s Activation Reagent is water-based and replaces the dry chemicals you would use with the traditional NHS/EDC method. Our One-Step-Activation only takes one hour, and improves efficiency in terms of both time and cost. It also provides the ability to either store activated particles up to 12 months for later use, or to immediately couple proteins.

Application

Particle-Based Immunoassays, Lateral Flow, Bioseparations and Immunoprecipitation

Features and Benefits

  • For activation of carboxylated magnetic and non-magnetic particles from 200 nm to 3 μm
  • Built upon patented technology using chelation and coordination chemistry
  • Flexibility to couple antibodies, antigens and problematic proteins
  • Reduce reagent preparation time by 3-4 hours
  • Reduced aggregation

Other Notes

  • Particles: Not included in this kit. This kit is for magnetic and latex particles
  • For Best Results: You will require a sonication bath to use this kit
  • Base: Water-based Activation Reagent is included in this Kit

Kit Components Only

Product No.
Description

  • Particle Pretreatment Solution

  • Particle Activation Solution

  • Particle Intermediate Solution

  • Coupling Buffer, Formulation C

  • Blocking Buffer, Formulation B

  • Storage Buffer, Formulation A

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service