Skip to Content
Merck
All Photos(7)

Key Documents

SAB2701800

Sigma-Aldrich

Anti-sox2 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

cb236, sox2, wu:fb83g04, wu:fc14d07

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

zebrafish, (Japanese Medaka)

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... sox2(378723)

Immunogen

Recombinant protein containing a sequence corresponding to a region within amino acids 42 and 238 of sox2 according to NP_998283

Application

Suggested starting dilutions are as follows: IHC-Fr: 1:100-1:1000, IHC-P: Assay-dependent dilution, WB: Assay-dependent dilution, IHC-Wm: 1:50-1:500. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Biochem/physiol Actions

Transcriptional activator. May function as a switch in neuronal development.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

1XPBS, 20% Glycerol (pH7). 0.025% ProClin 300 was added as a preservative.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jolien Van Houcke et al.
NPJ Regenerative medicine, 8(1), 31-31 (2023-06-17)
The young African turquoise killifish has a high regenerative capacity, but loses it with advancing age, adopting several aspects of the limited form of mammalian regeneration. We deployed a proteomic strategy to identify pathways that underpin the loss of regenerative
Jolien Van Houcke et al.
Aging cell, 20(9), e13464-e13464 (2021-08-25)
The aging central nervous system (CNS) of mammals displays progressive limited regenerative abilities. Recovery after loss of neurons is extremely restricted in the aged brain. Many research models fall short in recapitulating mammalian aging hallmarks or have an impractically long
Sophie Vanhunsel et al.
NPJ aging and mechanisms of disease, 7(1), 22-22 (2021-08-19)
Worldwide, people are getting older, and this prolonged lifespan unfortunately also results in an increased prevalence of age-related neurodegenerative diseases, contributing to a diminished life quality of elderly. Age-associated neuropathies typically include diseases leading to dementia (Alzheimer's and Parkinson's disease)
Kyeong-Hye Moon et al.
eLife, 9 (2021-01-01)
Defective primary cilia cause a range of diseases known as ciliopathies, including hearing loss. The etiology of hearing loss in ciliopathies, however, remains unclear. We analyzed cochleae from three ciliopathy mouse models exhibiting different ciliogenesis defects: Intraflagellar transport 88 (Ift88)
Ana Benito-Gonzalez et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(38), 12865-12876 (2014-09-19)
Mechano-sensory hair cells (HCs), housed in the inner ear cochlea, are critical for the perception of sound. In the mammalian cochlea, differentiation of HCs occurs in a striking basal-to-apical and medial-to-lateral gradient, which is thought to ensure correct patterning and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service