Skip to Content
Merck
All Photos(1)

Key Documents

FCMAB112F

Sigma-Aldrich

Anti-SOX-2 Antibody, clone 6G1.2, FITC conjugate

clone 6G1.2, from mouse, FITC conjugate

Synonym(s):

SRY (sex determining region Y)-box 2, SRY-related HMG-box gene 2, sex-determining region Y-box 2, transcription factor SOX2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

conjugate

FITC conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6G1.2, monoclonal

species reactivity

human, rat, mouse

technique(s)

flow cytometry: suitable

isotype

IgG2b

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SOX2(6657)
mouse ... Sox2(20674)
rat ... Sox2(499593)

Related Categories

General description

Sox-2 plays a key role in embryonic stem cell development, and has been shown to help stem cells maintain a pluripotent potential. It has also been shown to be associated with uncommitted dividing stem cell and precursor cells of the developing CNS.

Specificity

SOX-2

Application

Anti-SOX-2 Antibody, clone 6G1.2, FITC conjugate is an antibody against SOX-2 for use in FC.

Quality

Evaluated by flow cytometry on undifferentiated adult Rat hippocampal neural stem cells SCR022.
Optimal working dilutions must be determined by the end user.

Target description

~34 kDa

Physical form

Purified mouse monoclonal IgG2b conjugated to FITC in PBS with less than 0.09% sodium azide and 15 mg/mL BSA.

Analysis Note

Control
Rat Neural Stem Cells.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Breanna S Borys et al.
Biotechnology and bioengineering, 117(5), 1316-1328 (2020-01-22)
Embryonic stem cells (ESCs) have almost unlimited proliferation capacity in vitro and can retain the ability to contribute to all cell lineages, making them an ideal platform material for cell-based therapies. ESCs are traditionally cultured in static flasks on a
Thalia Jimenez et al.
American journal of cancer research, 10(10), 3487-3506 (2020-11-10)
Breast cancer remains a complex disease resulting in high mortality in women. A subset of cancer stem cell (CSC)-like cells expressing aldehyde dehydrogenase 1 (ALDH1) and SOX2/OCT4 are implicated in aggressive biology of specific subtypes of breast cancer. Targeting these

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service