Skip to Content
Merck
All Photos(3)

Documents

SAB2701049

Sigma-Aldrich

Anti-CDT1 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

CDT1, DUP, RIS2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry: suitable
indirect immunofluorescence: suitable
western blot: 500-3000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CDT1(81620)

General description

Chromatin licensing and DNA replication factor 1/Cdc10-dependent transcript 1 (CDT1), a G1-specific cell-cycle factor, belongs to the pre-replication complex (pre-RC). The CDT1 gene is located on the human chromosome 16q24.3.

Immunogen

Synthetic peptide containing a sequence corresponding to a region within amino acids 448 and 490 (Q9H211) of CDT1 according to AAH08676

Application

Suggested starting dilutions are as follows: ICC/IF: 1:100-1:1000, IHC-P: 1:100-1:1000, WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Biochem/physiol Actions

Chromatin licensing and DNA replication factor 1/Cdc10-dependent transcript 1 (CDT1) is involved in minichromosome maintenance (MCM) loading at the G1 phase of the cell cycle. It plays a key role in origin licensing in eukaryotic cells. It is a DNA replication protein that stimulates the production of pre-replicative complexes and replication in the next cell cycle by accumulating at the end of the cell cycle. The CDT1 gene mutation can cause Meier–Gorlin syndrome, a type of primordial dwarfism.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

1XPBS, 20% Glycerol (pH7). 0.025% ProClin 300 was added as a preservative.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrea Ballabeni et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(30), E2848-E2853 (2013-07-10)
Cdc10-dependent transcript 1 (Cdt1) is an essential DNA replication protein whose accumulation at the end of the cell cycle promotes the formation of pre-replicative complexes and replication in the next cell cycle. Geminin is thought to be involved in licensing
Georgia Xouri et al.
Cell cycle (Georgetown, Tex.), 6(13), 1549-1552 (2007-06-30)
Within each cell cycle, a cell must ensure that the processes of selection of replication origins (licensing) and initiation of DNA replication are well coordinated to prevent reinitiation of DNA replication from the same DNA segment during the same cell
Vassilis Roukos et al.
Journal of cell science, 124(Pt 3), 422-434 (2011-01-13)
For genomic integrity to be maintained, the cell cycle and DNA damage responses must be linked. Cdt1, a G1-specific cell-cycle factor, is targeted for proteolysis by the Cul4-Ddb1(Cdt2) ubiquitin ligase following DNA damage. Using a laser nanosurgery microscope to generate
Pedro N Pozo et al.
Molecular biology of the cell, mbcE18040242-mbcE18040242 (2018-10-04)
The earliest step in DNA replication is origin licensing which is the DNA loading of MCM helicase complexes. The Cdt1 protein is essential for MCM loading during G1 phase of the cell cycle, yet the mechanism of Cdt1 function is
Hui-Hui Xie et al.
Medicine, 100(7), e24382-e24382 (2021-02-21)
Subchromosomal deletions and duplications could currently be detected by noninvasive preliminary screening (NIPS). However, NIPS is a screening test that requires further diagnosis. Here we report a fetus with an autosomal abnormality revealed by NIPS and conventional karyotype combined with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service