Skip to Content
Merck
All Photos(2)

Key Documents

SAB2500736

Sigma-Aldrich

Anti-P70S6K/RPS6KB1 antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-PS6K, Anti-Ribosomal protein S6 kinase

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, rabbit, human, mouse

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RPS6KB1(6198)

Immunogen

Peptide with sequence C-MISKRPEHLRMNL from the C Terminus of the protein sequence according to NP_003152.1.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guanli Zhang et al.
Journal of biochemical and molecular toxicology, 37(1), e23237-e23237 (2022-10-05)
Pre-eclampsia (PE) is a major cause of hypertension in maternal and fetal. Atlastin-1 (ATL1), one regulator of endoplasmic reticulum (ER) morphology, participates in tubular ER formation and protein synthesis. The objective of this study is to investigate the role and
Hui-Ying Huang et al.
Translational cancer research, 11(5), 1076-1088 (2022-06-17)
New and effective chemotherapy or targeted therapy strategies are needed against laryngeal squamous cell carcinoma (LSCC). We aimed to explore the antitumor effect of dual PI3K/mTOR inhibitor combined with autophagy suppression on LSCC and its underlying mechanism. Hep-2 and AMC-HN-8

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service