Skip to Content
Merck
All Photos(1)

Key Documents

SAB1306410

Sigma-Aldrich

ANTI-NIK (CENTER) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

MAP3K14, Mitogen-activated protein kinase kinase kinase 14, NF-kappa-beta-inducing kinase, NIK, Serine/threonine-protein kinase NIK

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

104042 Da

species reactivity

human, rat

technique(s)

immunohistochemistry: 1:50-1:100
western blot: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... MAP3K14(9020)

Physical form

Supplied in PBS with 0.09% (W/V) sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xander M R van Wijk et al.
The Biochemical journal, 461(3), 461-468 (2014-05-14)
HS (heparan sulfate) is a long linear polysaccharide, variably modified by epimerization and sulfation reactions, and is organized into different domains defined by the extent of modification. To further elucidate HS structural organization, the relative position of different HS structures
Angelique L W M M Rops et al.
Kidney international, 86(5), 932-942 (2014-04-25)
The glomerular endothelial glycocalyx is postulated to be an important modulator of permeability and inflammation. The glycocalyx consists of complex polysaccharides, the main functional constituent of which, heparan sulfate (HS), is synthesized and modified by multiple enzymes. The N-deacetylase-N-sulfotransferase (Ndst)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service