Merck
  • 'Immunosequencing' of heparan sulfate from human cell lines and rat kidney: the (GlcNS6S-IdoA2S)₃ motif, recognized by antibody NS4F5, is located towards the non-reducing end.
'Immunosequencing' of heparan sulfate from human cell lines and rat kidney: the (GlcNS6S-IdoA2S)₃ motif, recognized by antibody NS4F5, is located towards the non-reducing end.
The Biochemical journal (2014-05-14)
Xander M R van Wijk, Arie Oosterhof, Elly M M Versteeg, Els M A van de Westerlo, Toin H van Kuppevelt
ABSTRACT
HS (heparan sulfate) is a long linear polysaccharide, variably modified by epimerization and sulfation reactions, and is organized into different domains defined by the extent of modification. To further elucidate HS structural organization, the relative position of different HS structures, identified by a set of phage-display-derived anti-HS antibodies, was established. Two strategies were employed: inhibition of HS biosynthesis using 4-deoxy-GlcNAc, followed by resynthesis, and limited degradation of HS using heparinases. Using both approaches, information about the position of antibody-defined HS structures was identified. The HS structure recognized by the antibody NS4F5, rigorously identified as (GlcN6S-IdoA2S)₃, was found towards the non-reducing end of the HS chain.
Sigma-Aldrich
2-Deoxy-D-glucose, ≥98% (GC), BioXtra
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
2-Deoxy-D-glucose, ≥98% (GC), crystalline
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
2-Deoxy-D-glucose, ≥99% (GC), crystalline