Skip to Content
Merck
All Photos(1)

Key Documents

S7448

Sigma-Aldrich

STF-62247

≥98% (HPLC), solid

Synonym(s):

N-(3-Methylphenyl)-4-(4-pyridinyl)-2-thiazolamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H13N3S
CAS Number:
Molecular Weight:
267.35
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

solubility

DMSO: soluble 26 mg/mL
H2O: insoluble

storage temp.

room temp

SMILES string

Cc1cccc(Nc2nc(cs2)-c3ccncc3)c1

InChI

1S/C15H13N3S/c1-11-3-2-4-13(9-11)17-15-18-14(10-19-15)12-5-7-16-8-6-12/h2-10H,1H3,(H,17,18)

InChI key

KATNUHQNJGNLPW-UHFFFAOYSA-N

Application

STF-62247 may be used as positive control in autophagy inducing cell signaling studies.

Biochem/physiol Actions

STF-62247 induces the formation of large vacuoles in the von Hippel-Lindau deficient renal carcinoma cells, a characteristic of autophagy.1
STF-62247 is a selective inducer of autophagy in VHL-deficient renal cell carcinoma cells.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sandra Turcotte et al.
Autophagy, 4(7), 944-946 (2008-09-05)
Radiation and conventional cytotoxic chemotherapies are ineffective in treating renal cancer. Approximately 75 percent of renal cell carcinoma (RCC) is associated with an inactivation of the tumor suppressor gene von Hippel-Lindau (VHL). We exploited the possibility of targeting VHL-deficient RCC
Tomohiro Kozako et al.
BMC cancer, 18(1), 791-791 (2018-08-08)
Sirtuin 2 (SIRT2) is a member of the sirtuin family, nicotinamide adenine dinucleotide+-dependent deacylases, which participates in modulation of cell cycle control, neurodegeneration, and tumorigenesis. SIRT2 expression increases in acute myeloid leukemia blasts. Downregulation of SIRT2 using siRNA causes apoptosis
Kanchan Phadwal et al.
Autophagy, 8(4), 677-689 (2012-02-04)
Autophagy is a conserved constitutive cellular process, responsible for the degradation of dysfunctional proteins and organelles. Autophagy plays a role in many diseases such as neurodegeneration and cancer; however, to date, conventional autophagy detection techniques are not suitable for clinical
Maximilian N Kinzler et al.
Scientific reports, 10(1), 687-687 (2020-01-22)
Induction of autophagy can have beneficial effects in several human diseases, e.g. cancer and neurodegenerative diseases (ND). Here, we therefore evaluated the potential of two novel autophagy-inducing compounds, i.e. STF-62247 and pimozide, to stimulate autophagy as well as autophagic cell
Nadia Bouhamdani et al.
Carcinogenesis, 41(6), 828-840 (2019-09-27)
The inactivation of the tumor suppressor gene, von Hippel-Lindau (VHL), has been identified as the earliest event in renal cell carcinoma (RCC) development. The loss of heterogeneity by chromosome 3p deletion followed by inactivating mutations on the second VHL copy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service