Skip to Content
Merck
All Photos(1)

Key Documents

S5637

Sigma-Aldrich

Sulfamethazine sodium salt

≥98%

Synonym(s):

4-Amino-N-(4,6-dimethyl-2-pyrimidinyl)benzenesulfonamide

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$64.50
100 G
$93.30

$64.50

PRICE WITHOUT NATIONAL TAXES

1 left in stockDetails

Request a Bulk Order

Select a Size

Change View
25 G
$64.50
100 G
$93.30

About This Item

Linear Formula:
C12H13N4O2SNa
CAS Number:
Molecular Weight:
300.31
EC Number:
MDL number:
UNSPSC Code:
51284910
PubChem Substance ID:
NACRES:
NA.85

$64.50

PRICE WITHOUT NATIONAL TAXES

1 left in stockDetails

Request a Bulk Order

Quality Level

Assay

≥98%

form

powder

storage condition

(Keep container tightly closed in a dry and well-ventilated place.)

color

white to off-white

solubility

H2O: soluble 50 mg/mL

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

DNA synthesis | interferes
enzyme | inhibits

storage temp.

2-8°C

SMILES string

[Na].Cc1cc(C)nc(NS(=O)(=O)c2ccc(N)cc2)n1

InChI

1S/C12H14N4O2S.Na.H/c1-8-7-9(2)15-12(14-8)16-19(17,18)11-5-3-10(13)4-6-11;;/h3-7H,13H2,1-2H3,(H,14,15,16);;

InChI key

WIVZAHIZHZEEOX-UHFFFAOYSA-N

General description

Chemical structure: sulfonamide

Application

Sulfamethazine is an antibiotic used to clinically treat bronchitis, prostatitis and urinary tract infections[1]. It is used in disposition and depletion kinetic studies[2][3]. It is used to develop detection techniques for quantification in fluids such as cows′ milk, honey and swine urine[4].

Biochem/physiol Actions

Sulfamethazine is an antimicrobial sulfur drug that blocks the synthesis of dihydrofolic acid by inhibiting the enzyme dihydropteroate synthase. Sulfamethazine is a competitive inhibitor of bacterial para-aminobenzoic acid (PABA), which is required for bacterial synthesis of folic acid[1]. It induces CYP3A4 expression and is acetylated by N-acetyltransferase. It exhibits sex dependent pharmacokinetics, metabolized by the male specific isoform CYP2C11. Sulfamethazine is bacteriostatic.

Packaging

25G,100G

Other Notes

Keep container tightly closed in a dry and well-ventilated place.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A D Mitchell et al.
Drug metabolism and disposition: the biological fate of chemicals, 14(2), 161-165 (1986-03-01)
Swine weighing 60-70 kg were orally administered 14C-sulfamethazine [4-amino-N-(4,6-dimethyl-2-pyrimidinyl)benzene[U-14C]sulfonamide] at 12-hr intervals for 7 days (165 mg/dose; 0.126-5.04 mCi/mmol). The animals were sacrificed at 8 hr or 2, 5, or 10 days after the last dose was given and tissues
Bożena Czubat et al.
Frontiers in microbiology, 11, 2008-2008 (2020-09-26)
MSMEG_4305 is a two-domain protein of Mycolicibacterium smegmatis (Mycobacterium smegmatis) (Mycolicibacterium smegmatis). The N-terminal domain of MSMEG_4305 encodes an RNase H type I. The C-terminal domain is a presumed CobC, predicted to be involved in the aerobic synthesis of vitamin
Tangbin Yang et al.
Hybridoma (2005), 29(5), 403-407 (2010-11-06)
A specific monoclonal antibody (MAb) against sulfamethazine was produced with hybridoma technology. This assay shows very high sensitivity with IC50 of 0.4 ng/mL and LOD of 0.05 ng/mL when it was run in 0.02 mol/L PBS (pH 7.5). This MAb has shown high
C J Chapron et al.
Journal of clinical pharmacology, 16(7), 338-344 (1976-07-01)
The relationship between sulfamethazine disposition kinetics and acetylation phenotype was studied in man. Sulfamethazine pharmacokinetic parameters were determined after the administration of the drug as an oral suspension. When the half-life, acetylation rate constant, or per cent available dose excreted
Craig Knox et al.
Nucleic acids research, 39(Database issue), D1035-D1041 (2010-11-10)
DrugBank (http://www.drugbank.ca) is a richly annotated database of drug and drug target information. It contains extensive data on the nomenclature, ontology, chemistry, structure, function, action, pharmacology, pharmacokinetics, metabolism and pharmaceutical properties of both small molecule and large molecule (biotech) drugs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service