Skip to Content
Merck
All Photos(2)

Key Documents

S0800

Sigma-Aldrich

Sulfamerazine sodium salt

Synonym(s):

4-Amino-N-(4-methyl-2-pyrimidinyl)benzenesulfonamide

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$206.00

$206.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Select a Size

Change View
250 G
$206.00

About This Item

Linear Formula:
C11H11N4O2SNa
CAS Number:
Molecular Weight:
286.29
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$206.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

SMILES string

[Na+].Cc1ccnc([N-]S(=O)(=O)c2ccc(N)cc2)n1

InChI

1S/C11H11N4O2S.Na/c1-8-6-7-13-11(14-8)15-18(16,17)10-4-2-9(12)3-5-10;/h2-7H,12H2,1H3;/q-1;+1

InChI key

BSFJGCCAXDCMOX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Involved in characterization of chirosan membranes for the transport of drugs

Quality

Contains residual methanol

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service