Skip to Content
Merck
Sucrose British Pharmacopoeia (BP) Reference Standard

Sucrose

Sucrose British Pharmacopoeia (BP) Reference Standard
Synonym(s):
α-D-Glc-(1→2)-β-D-Fru, α-D-Glucopyranosyl β-D-fructofuranoside, β-D-Fructofuranosyl-α-D-glucopyranoside, D(+)-Saccharose, Sugar, Sucrose
Empirical Formula (Hill Notation):
C12H22O11
CAS Number:
Molecular Weight:
342.30
EC Number:

Products

Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Format
Research Area
Quality Segment
Application
Optical Isomer
Reference Material Type
Special Grade
Technique
Showing 1-20 of 25
Page 1 of 2
Page 1 of 2