Skip to Content
Merck
All Photos(2)

Documents

S0383

Sigma-Aldrich

Sulfameter

Synonym(s):

5-Methoxysulfadiazine, N1-(5-Methoxypyrimidin-2-yl)sulfanilamide, Sulfamethoxydiazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12N4O3S
CAS Number:
Molecular Weight:
280.30
Beilstein:
621130
EC Number:
MDL number:
UNSPSC Code:
51283913
PubChem Substance ID:
NACRES:
NA.85

form

powder

storage condition

(Keep container tightly closed in a dry and well-ventilated place. Light sensitive.)

color

white to faint yellow

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria
mycobacteria
parasites

Mode of action

enzyme | inhibits

storage temp.

2-8°C

SMILES string

COc1cnc(NS(=O)(=O)c2ccc(N)cc2)nc1

InChI

1S/C11H12N4O3S/c1-18-9-6-13-11(14-7-9)15-19(16,17)10-4-2-8(12)3-5-10/h2-7H,12H2,1H3,(H,13,14,15)

InChI key

GPTONYMQFTZPKC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Chemical structure: sulfonamide

Application

Sulfameter is a sulfonamide antibiotic used as a leprostatic agent and to treat urinary tract infections. It has been investigated as a potential therapy for chloroquine-resistant malaria during pregnancy.

Biochem/physiol Actions

Sulfameter is a sulfonamide antibiotic. Sulfonamides block the synthesis of dihydrofolic acid by inhibiting the enzyme dihydropteroate synthase. Sulfonamides are competitive inhibitors of bacterial para-aminobenzoic acid (PABA), which is required for bacterial synthesis of folic acid. Sulfameter is a dihydrofolate reductase (DHFR) inhibitor. Sulfonamides are active against Gram positive bacteria and Gram negative bacteria. Mode of resistance is via the alteration of dihydropteroate synthase or alternative pathway for folic acid synthesis.

Other Notes

Keep container tightly closed in a dry and well-ventilated place. Light sensitive.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Forland et al.
Diabetes care, 8(5), 499-506 (1985-09-01)
Forty-five women with diabetes mellitus and urinary tract infections have been followed an average of 34 mo on treatment protocols based on localization of infection as determined by the presence or absence of antibody-coated bacteria (ACB). Treatment was usually, but
Treatment of Chloroquine-Resistant Malaria During Pregnancy
Elliott K. Main, Denise M. Main, et al.
Journal of the American Medical Association, 249, 3207-3209 (1983)
H S Kaumeier
Arzneimittel-Forschung, 30(10), 1794-1800 (1980-01-01)
It is demonstrated that the bioavailability of sulfamethoxydiazine (Durenat), taken as an example, is much better when administered simultaneously with a test meal than when given in the fasting state. Also a high-lipid meal in comparison with a high-protein or
Identification of polymorphs of sulfamethoxydiazine by optical and morphological methods.
T Yokoyama et al.
Chemical & pharmaceutical bulletin, 26(4), 1044-1048 (1978-04-01)
The effect of gastro-intestinal filling on the absorption of sulfametoxydiazine.
S Kaumeier et al.
International journal of clinical pharmacology and biopharmacy, 17(10), 412-415 (1979-10-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service