Skip to Content
Merck
All Photos(1)

Key Documents

RB45

Sigma-Aldrich

Reactive Blue 4−Agarose

prepacked column

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
23151817
PubChem Substance ID:
NACRES:
NA.56

form

prepacked column

column volume

2.5 mL

storage temp.

2-8°C

SMILES string

O=C1C2=C(C(NC3=CC=C(S(=O)(O)=O)C(NC4=NC(Cl)=NC(Cl)=N4)=C3)=CC(S(=O)(O)=O)=C2N)C(C5=C1C=CC=C5)=O

InChI

1S/C23H14Cl2N6O8S2/c24-21-29-22(25)31-23(30-21)28-12-7-9(5-6-14(12)40(34,35)36)27-13-8-15(41(37,38)39)18(26)17-16(13)19(32)10-3-1-2-4-11(10)20(17)33/h1-8,27H,26H2,(H,34,35,36)(H,37,38,39)(H,28,29,30,31)

InChI key

RTLULCVBFCRQKI-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Reactive Blue 4-agarose is used in affinity chromatography, protein chromatography and dye resins. Reactive Blue 4-agarose has been used to express human pituitary glutaminyl cyclase (hQC) in Drosophila S2 cells.

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Kit Components Also Available Separately

Product No.
Description
SDS

  • R2507Reactive Blue 4−Agarose, saline suspension 5 columnSDS

Storage Class Code

12 - Non Combustible Liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D E Cane et al.
Archives of biochemistry and biophysics, 304(2), 415-419 (1993-08-01)
Aristolochene synthase, a fungal cyclase which has been isolated from Aspergillus terreus and Penicillium roqueforti, catalyzes the cyclization of farnesyl diphosphate to the sesquiterpene hydrocarbon aristolochene. The aristolochene synthase gene (Ari1) of P. roqueforti has previously been cloned and expressed
Rachell E Booth et al.
Protein expression and purification, 32(1), 141-146 (2003-12-19)
Human pituitary glutaminyl cyclase (hQC) was expressed in Drosophila S2 cells under the control of an inducible metallothionene promoter and fused to the Drosophila immunoglobulin-binding protein signal sequence to enable secretion into the culture media. Expression levels reached 50 microg/mL

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service