Skip to Content
Merck
All Photos(2)

Documents

RAB0177

Sigma-Aldrich

Human Fas ELISA Kit

for cell and tissue lysates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type cell lysate
sample type tissue lysate

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 5 pg/mL
standard curve range: 2.74-2000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... FAS(14102)

General description

The Human Fas ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human Fas cell lysate and tissue lysate.

Immunogen

Recombinant Human Fas

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABDIL6ELISA 5X Sample Diluent Buffer (Item D2)SDS

  • RABELADEELISA 5X Assay/Sample Diluent Buffer E (Item E2)SDS

  • RABLYSIS12X Cell Lysis Buffer (Item J)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ignatios Ikonomidis et al.
Circulation. Cardiovascular imaging, 7(4), 619-628 (2014-05-02)
We investigated the effects of anakinra, an interleukin-1 receptor antagonist, on coronary and left ventricular function in coronary artery disease (CAD) patients with rheumatoid arthritis. In a double-blind crossover trial, 80 patients with rheumatoid arthritis (60 with CAD and 20
Mark T Kearns et al.
American journal of physiology. Lung cellular and molecular physiology, 307(1), L62-L70 (2014-05-20)
Apoptosis of alveolar macrophages and their subsequent clearance by neighboring phagocytes are necessary steps in the resolution of acute pulmonary inflammation. We have recently identified that activation of the Fas death receptor on the cell surface of macrophages drives macrophage
Anil K Taneja et al.
International journal of cardiology, 175(2), 253-260 (2014-06-01)
Heart failure activates neurohormones, and elevated levels of brain natriuretic peptide (BNP) are associated with adverse outcomes. The SENIORS trial showed that nebivolol, a highly selective beta-1 antagonist with vasodilating properties, reduced the composite outcome of all cause mortality or
K Mizrahi et al.
Bone marrow transplantation, 49(7), 942-949 (2014-04-30)
The influence of TNF-α and Fas-ligand (FasL) on viability and function was evaluated in fresh- and expanded-umbilical cord blood (UCB) cells. CD34(+) progenitors and T cells display outstanding survival, whereas ~30% and >50% B lymphocytes and myeloid cells undergo spontaneous
Soon Woo Nam et al.
Clinical science (London, England : 1979), 127(8), 519-526 (2014-04-10)
Cirrhotic cardiomyopathy is defined as systolic and diastolic dysfunctions, electrophysiological changes and macroscopic structural changes. However, the underlying mechanisms of this syndrome remain unclear. A possible role of myocardial apoptosis in the pathogenesis has not been previously examined. We hypothesized

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service