Skip to Content
Merck
All Photos(1)

Key Documents

RABLYSIS1

Sigma-Aldrich

2X Cell Lysis Buffer (Item J)

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$38.50

$38.50

PRICE WITHOUT NATIONAL TAXES

Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
5 ML
$38.50

About This Item

UNSPSC Code:
12161703
NACRES:
NA.32

$38.50

PRICE WITHOUT NATIONAL TAXES

Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

shipped in

wet ice

storage temp.

−20°C

Other Notes

Intended for use with ELISA kits.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhongbai Zhang et al.
Experimental and therapeutic medicine, 21(6), 556-556 (2021-04-15)
Ischemia-reperfusion (I/R) plays an important role in myocardial damage, which has been widely recognized as a key procedure in the cardiovascular disease. A hypoxia/reoxygenation (H/R) model was established using H9c2 cardiomyocytes to investigate the possible positive effect of oxymatrine (OMT)
Liqun Chu et al.
International journal of molecular medicine, 44(4), 1377-1387 (2019-08-23)
The retina is sensitive to injury resulting from oxidative stress (OS) due to its high oxygen consumption. Patients with retinitis pigmentosa suffer from excessive OS. N‑acetylcysteine (NAC) is used as a mucolytic agent for the clinical treatment of disorders, such

Questions

  1. How long is the stability date of this product?

    1 answer
    1. This product can be stored at 4°C for 6 months or frozen at -80°C for 1 year.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service