RAB0090
Human CEACAM-1 ELISA Kit
for serum, plasma, cell culture supernatant and urine
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41116158
NACRES:
NA.32
Recommended Products
species reactivity
human
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
capture ELISA: suitable
input
sample type urine
sample type serum
sample type cell culture supernatant(s)
sample type plasma
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 20 pg/mL
standard curve range: 20.58-15000 pg/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... CEACAM1(634)
Related Categories
General description
The Human CEACAM-1 (Carcinoembryonic Antigen- related Cell Adhesion Molecule 1) ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human CEACAM-1 in serum, plasma, cell culture supernatants and urine.
Serum carcinoembryonic antigen-related cell adhesion molecule 1 (CEACAM1) is a transmembrane glycoprotein and is localized to endothelial, epithelial, lymphoid and myeloid cells.The CEACAM-1 gene is mapped to human chromosome 19q13.2.
Serum carcinoembryonic antigen-related cell adhesion molecule 1 (CEACAM1) is a transmembrane glycoprotein and is localized to endothelial, epithelial, lymphoid and myeloid cells.The CEACAM-1 gene is mapped to human chromosome 19q13.2.
Immunogen
Recombinant Human CEACAM1
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Biochem/physiol Actions
Serum carcinoembryonic antigen-related cell adhesion molecule 1 (CEACAM1) participates in cell adhesion mechanism through homophilic or heterophilic interactions. Abnormal expression of the CEACAM1 gene is observed in a number of tumors. Ceacam1 possesses tyrosine phosphatase, an inhibitory motif, via which the receptor activates its inhibitory signals.
Other Notes
A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.
Please type the word sample in the text box provided for lot number.
Kit Components Also Available Separately
Product No.
Description
SDS
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
SOX9 indirectly regulates CEACAM1 expression and immune resistance in melanoma cells.
Ashkenazi S, et al.
Oncotarget, 7(21), 30166-30166 (2016)
Rong Zhang et al.
Journal of ethnopharmacology, 155(1), 104-112 (2014-05-03)
The present study systematically investigate the in vivo and in vitro effect of total lignans (TL) extracted from Eucommia ulmoides Oliv. barks on bone formation using ovariectomy rat model and primary cultures of rat osteoblasts. Eighty 3-month-old female Sprague-Dawley rats
Suppression of human metapneumovirus (HMPV) infection by the innate sensing gene CEACAM1.
Diab M, et al.
Oncotarget, 7(41), 66468-66468 (2016)
Up-regulation of carcinoembryonic antigen-related cell adhesion molecule 1 in gastrointestinal cancer and its clinical relevance.
Zhou M, et al.
Acta biochimica et biophysica Sinica, 49(8), 737-743 (2017)
CEACAM1: contact-dependent control of immunity.
Gray-Owen S D and Blumberg R S
Nature Reviews: Immunology, 6(6), 433-433 (2006)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service