Skip to Content
Merck
All Photos(1)

Key Documents

R4526

Sigma-Aldrich

Reference Dye for Quantitative PCR

100 ×, solution

Synonym(s):

Reference dye for qPCR, ROX

Sign Into View Organizational & Contract Pricing

Select a Size

0.3 ML
$22.70
5 ML
$342.00

$22.70

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025



Select a Size

Change View
0.3 ML
$22.70
5 ML
$342.00

About This Item

UNSPSC Code:
41106300
NACRES:
NA.55

$22.70

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


form

solution

usage

sufficient for ≥600 reactions

concentration

100 ×

technique(s)

PCR: suitable

color

red

solubility

water: soluble

storage temp.

2-8°C

General description

Sigma′s Reference Dye for quantitative polymerase chain reaction (qPCR) is a proprietary dye for use with real-time PCR. It is used for the normalization of reaction data when using SYBR Green, molecular probes, or dual-labeled probe chemistries for real-time detection. The Reference Dye is supplied as a 100× solution with a maximum excitation and emission of 586 nm and 605 nm, respectively. Instrument settings for ROX reference dye are satisfactory for the measurement of Reference Dye for qPCR.

Application

Reference Dye for Quantitative PCR has been used:
  • in the preparation of mastermix for real time-quantitative polymerase chain reaction (RT-qPCR)[1][2]
  • as a component of the reaction mixture for detection of Clostridium difficile by quantitative polymerase chain reaction (qPCR)[3][4]
  • for analyzing the degree of cellular DNA contamination by qPCR[5]

Other Notes

Reference Dye for Quantitative PCR is forR&D use only. Not for drug, household, or other uses.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sambrook, J. et al.
Molecular Cloning: A Laboratory Manual, 3rd (2000)
Araceli Melendez-Avalos et al.
Folia microbiologica, 65(1), 133-142 (2019-05-20)
This study aimed to analyze the proinflammatory cytokine mRNA expression in the urinary tract of BALB/c mice infected with bacterial strains with uropathogenic potential. Groups of four 6-week-old female BALB/c mice were intraurethrally inoculated with 5 × 107 colony-forming units (CFU) of
Gustav Johansson et al.
Molecular cancer therapeutics, 20(12), 2568-2576 (2021-09-24)
The majority of patients diagnosed with advanced gastrointestinal stromal tumors (GISTs) are successfully treated with a combination of surgery and tyrosine kinase inhibitors (TKIs). However, it remains challenging to monitor treatment efficacy and identify relapse early. Here, we utilized a
Partial deletion of rng (RNase G)-enhanced homoethanol fermentation of xylose by the non-transgenic Escherichia coli RM10
Manow R, et al.
Journal of Industrial Microbiology & Biotechnology, 39(7), 977-985 (2012)
Increased environmental sample area and recovery of Clostridium difficile spores from hospital surfaces by quantitative PCR and enrichment culture
Brown KA, et al.
Infection Control and Hospital Epidemiology : The Official Journal of the Society of Hospital Epidemiologists of America, 39(8), 917-923 (2018)

Articles

Identify causes and remedies for SDS-PAGE sample preparation challenges and optimize electrophoresis conditions.

Protocols

Protocol describes amplification of DNA through quantitative PCR with SYBR Green. Consistent batch-to-batch performance can be achieved with large numbers of PCR reactions.

Our SYBR Green qPCR Protocol is a method designed to detect accurate quantification of gene expression and RT-PCR reactions

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service