Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.
Rat Skeletal Muscle Cells (RSkMC) provide a useful system to study many aspects of muscular function and disease and can serve as an intermediate model of human diseases. Skeletal Muscle Cells also play an instrumental role in the glucose metabolism and diabetes.
Cell Line Origin
Muscle
Application
lipid and glucose metabolism, exercise, insulin action, cell signaling
Components
Rat Skeletal Muscle Cell Basal Medium containing 10% FBS & 10% DMSO
Preparation Note
1st passage, >500,000 cells in Rat Skeletal Muscle Cell Basal Medium containing 10% FBS & 10% DMSO
Journal of cellular physiology, 234(11), 19675-19690 (2019-04-05)
Interleukin (IL)-8 is released both in visceral adipose tissue and in contracting skeletal muscles. In this study, we examined cellular pathways associated with muscle hypertrophy, chosen on the basis of microRNA profiling, in differentiating rat primary skeletal muscle cells (RSkMC)
Questions
Reviews
★★★★★ No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.