Skip to Content
Merck
All Photos(1)

Key Documents

870792P

Avanti

D-threo-PPMP

Avanti Research - A Croda Brand 870792P, powder

Synonym(s):

1R,2R-(+)-1-phenyl-2-palmitoylamino-3-N-morpholine-1-propanol

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$315.00

$315.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$315.00

About This Item

Empirical Formula (Hill Notation):
C29H50N2O3
CAS Number:
Molecular Weight:
474.72
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$315.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 5 mg (870792P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870792P

lipid type

bioactive lipids
sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@H](C1=CC=CC=C1)[C@H](NC(CCCCCCCCCCCCCCC)=O)CN2CCOCC2

InChI key

OFBANDBMHLEMFA-XRKRLSELSA-N

Application

D-threo-1-phenyl-2-palmitoylamino-3-morpholino-1-propanol (D-threo-PPMP) has been used as an internal standard for the generation of standard curve in high performance liquid chromatography.[1]

Biochem/physiol Actions

D-threo-PPMP, also known as 1R,2R-(+)-1-phenyl-2-palmitoylamino-3-N-morpholine-1-propanol, is a bioactive sphingolipid. It plays a vital role in regulation of ceramide metabolism.[2] D-threo-PPMP influences cytokinesis failure[3] and glycosylation[2] by inhibiting glucosyl ceramide synthase (GCS).[3] D-threo-PPMP also stops acylation reaction by inhibiting the activity of 1-O-acylceramide synthase (1-O-ACS).[2]

Packaging

5 mL Amber Glass Screw Cap Vial (870792P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P H O'Donnell et al.
Leukemia, 16(5), 902-910 (2002-05-03)
The retinoid, N-(4-hydroxyphenyl)retinamide (4-HPR), mediates p53-independent cytotoxicity and can increase reactive oxygen species and ceramide in solid tumor cell lines. We determined changes in ceramide and cytotoxicity upon treatment with 4-HPR (3-12 microM) in six human acute lymphoblastic leukemia (ALL)
G E Atilla-Gokcumen et al.
Journal of the American Chemical Society, 133(26), 10010-10013 (2011-06-15)
Although cells undergo dramatic shape changes during cytokinesis, the role of the plasma membrane and lipids is poorly understood. We report that inactivation of glucosyl ceramide synthase (GCS), either by RNAi or with the small molecule PPMP, causes failure of
Xiaqin Wu et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 837(1-2), 44-48 (2006-05-24)
A high-performance liquid chromatography (HPLC) method was developed to measure levels of d-threo-1-phenyl-2-palmitoylamino-3-morpholino-1-propanol (d-threo-PPMP) in mouse plasma and liver. d-threo-PPMP was measured by HPLC with a Luna Pheny-Hexyl column (5 microm, 250 mm x 4.6 mm) employing UV detection at
E I de Chaves et al.
The Journal of biological chemistry, 272(5), 3028-3035 (1997-01-31)
Sphingolipids are abundant constituents of neuronal membranes and have been implicated in intracellular signaling. We show that two analogs of glycosphingolipid biosynthetic intermediates, fumonisin B1 (which inhibits dihydroceramide synthesis) and DL-1-phenyl-2-palmitoylamino-3-morpholino-1-propanol (PPMP) (which inhibits glucosylceramide synthesis) decrease glycosphingolipid synthesis in
Valérie Gouazé et al.
Cancer research, 65(9), 3861-3867 (2005-05-04)
Overexpression of glucosylceramide synthase (GCS), a pivotal enzyme in glycolipid biosynthesis, contributes to cancer cell resistance to chemotherapy. We previously showed that transfection of doxorubicin-resistant MCF-7-AdrR cells with GCS antisense restored cell sensitivity to doxorubicin and greatly enhanced sensitivity to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service