Skip to Content
Merck
All Photos(1)

Key Documents

PZ0321

Sigma-Aldrich

WYE-125132

≥97% (HPLC)

Synonym(s):

N-[4-[1-(1,4-Dioxaspiro[4.5]dec-8-yl)-4-(8-oxa-3-azabicyclo[3.2.1]oct-3-yl)-1H-pyrazolo[3,4-d]pyrimidin-6-yl]phenyl]-N′-methyl-urea, WYE-132

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$135.00
25 MG
$465.00

$135.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025



Select a Size

Change View
5 MG
$135.00
25 MG
$465.00

About This Item

Empirical Formula (Hill Notation):
C27H33N7O4
CAS Number:
Molecular Weight:
519.60
UNSPSC Code:
12352200
NACRES:
NA.77

$135.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Quality Level

Assay

≥97% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

N6(CC7OC(C6)CC7)c1nc(nc3[n](ncc31)C4CCC5(OCCO5)CC4)c2ccc(cc2)NC(=O)NC

InChI

1S/C27H33N7O4/c1-28-26(35)30-18-4-2-17(3-5-18)23-31-24(33-15-20-6-7-21(16-33)38-20)22-14-29-34(25(22)32-23)19-8-10-27(11-9-19)36-12-13-37-27/h2-5,14,19-21H,6-13,15-16H2,1H3,(H2,28,30,35)

InChI key

QLHHRYZMBGPBJG-UHFFFAOYSA-N

Biochem/physiol Actions

In ovarian cancer cells, WYE-132 might prevent the activity of sphingosine kinase-1 (SphK1), leading to pro-apoptotic ceramide production.[1]
WYE-125132 (WYE-132) is a highly potent, ATP-competitive, and specific inhibitor of mTOR with an IC50 value of 0.19 nM. Unlike rapamycin, which ihibits mTOR through allosteric binding to mTOR complex 1 (mTORC1) only, WYE-132 inhibits both mTORC1 and mTORC2. WYE-125132 is >5000-fold selective for mTOR vs. phosphoinositide 3-kinase (PI3K). WYE-132 induced apoptosis in a variety of cancer lines including LNCap prostate cells and H1975 lung cancer cells. WYE-132 also inhibited ovarian cancer cell growth.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Feiran Lu et al.
eLife, 4 (2015-12-10)
Niemann-Pick C1 (NPC1) is a lysosomal membrane protein that exports cholesterol derived from receptor-mediated uptake of LDL, and it also mediates cellular entry of Ebola virus. Cholesterol export is inhibited by nanomolar concentrations of U18666A, a cationic sterol. To identify
Dan Zhang et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 37(1), 1327-1336 (2015-08-22)
Epithelial ovarian cancer is the most common and lethal gynecological cancer in USA and around the world, causing major mortality annually. In the current study, we investigated the potential anti-ovarian cancer activity of WYE-132, a mammalian target of rapamycin (mTOR)
Iftah Marash et al.
Plant, cell & environment, 47(2), 629-650 (2023-10-31)
Plants constantly perceive and process environmental signals and balance between the energetic demands of growth and defense. Growth arrest upon pathogen attack was previously suggested to result from a redirection of the plants' metabolic resources towards the activation of plant
Sameer S Chopra et al.
Cell systems, 10(1), 66-81 (2019-12-10)
Frequent mutation of PI3K/AKT/mTOR signaling pathway genes in human cancers has stimulated large investments in targeted drugs but clinical successes are rare. As a result, many cancers with high PI3K pathway activity, such as triple-negative breast cancer (TNBC), are treated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service